Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228158383-228158620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJC2
Entrez Gene ID
57165
Gene Name
gap junction protein gamma 2
Gene Aliases
CX46.6, Cx47, GJA12, HLD2, LMPH1C, PMLDAR, SPG44
UniGene
Hs.100072
Gene Chromosome Location
Chr.1: 228149714-228159826 on Build GRCh38
Transcripts
NM_020435.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5114164 c.676G>A p.A226T Chr.1: 228158434-228158434 on Build GRCh38
3484435 c.789A>C p.E263D Chr.1: 228158547-228158547 on Build GRCh38
4827833 c.712G>A p.E238K Chr.1: 228158470-228158470 on Build GRCh38
7425727 c.707G>T p.G236V Chr.1: 228158465-228158465 on Build GRCh38
4028885 c.756C>T p.H252H Chr.1: 228158514-228158514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765995775 Chr.1: 228158586-228158586 on Build GRCh38
rs767386031 Chr.1: 228158552-228158552 on Build GRCh38
rs770175380 Chr.1: 228158499-228158499 on Build GRCh38
rs779717054 Chr.1: 228158428-228158428 on Build GRCh38
rs750135208 Chr.1: 228158424-228158424 on Build GRCh38
rs762995507 Chr.1: 228158522-228158522 on Build GRCh38
rs926523102 Chr.1: 228158505-228158505 on Build GRCh38
rs267606846 Chr.1: 228158536-228158536 on Build GRCh38
rs560832360 Chr.1: 228158493-228158493 on Build GRCh38
rs866103758 Chr.1: 228158534-228158534 on Build GRCh38
rs761638814 Chr.1: 228158409-228158409 on Build GRCh38
rs778181207 Chr.1: 228158470-228158470 on Build GRCh38
rs746163968 Chr.1: 228158490-228158490 on Build GRCh38
rs138851189 Chr.1: 228158560-228158560 on Build GRCh38
rs768752998 Chr.1: 228158523-228158523 on Build GRCh38
rs780983092 Chr.1: 228158482-228158483 on Build GRCh38
rs549807766 Chr.1: 228158538-228158538 on Build GRCh38
rs755777289 Chr.1: 228158427-228158427 on Build GRCh38
rs753309235 Chr.1: 228158436-228158436 on Build GRCh38
rs374172867 Chr.1: 228158575-228158575 on Build GRCh38
rs74315314 Chr.1: 228158572-228158572 on Build GRCh38
rs754510168 Chr.1: 228158595-228158595 on Build GRCh38
rs397514734 Chr.1: 228158545-228158545 on Build GRCh38
rs754488745 Chr.1: 228158463-228158463 on Build GRCh38
rs747898753 Chr.1: 228158564-228158566 on Build GRCh38
rs375623977 Chr.1: 228158474-228158474 on Build GRCh38
rs781697706 Chr.1: 228158488-228158488 on Build GRCh38
rs147024647 Chr.1: 228158407-228158407 on Build GRCh38
rs1057518455 Chr.1: 228158497-228158498 on Build GRCh38
rs761726866 Chr.1: 228158542-228158542 on Build GRCh38
rs753495849 Chr.1: 228158594-228158594 on Build GRCh38
rs74315313 Chr.1: 228158476-228158476 on Build GRCh38
rs796065029 Chr.1: 228158453-228158454 on Build GRCh38
rs529717425 Chr.1: 228158535-228158535 on Build GRCh38
rs760412672 Chr.1: 228158568-228158568 on Build GRCh38
rs771477313 Chr.1: 228158487-228158487 on Build GRCh38
rs775735957 Chr.1: 228158503-228158503 on Build GRCh38
rs767296387 Chr.1: 228158419-228158419 on Build GRCh38

View Full Product Details