Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 674611-674840 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEAF1
Entrez Gene ID
10522
Gene Name
DEAF1, transcription factor
Gene Aliases
MRD24, NUDR, SPN, ZMYND5
UniGene
Hs.243994
Gene Chromosome Location
Chr.11: 644220-695754 on Build GRCh38
Transcripts
NM_001293634.1 NM_021008.3 XM_011519842.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3452378 c.1284C>T p.V428V Chr.11: 674755-674755 on Build GRCh38
6642100 c.1351C>T p.R451W Chr.11: 674688-674688 on Build GRCh38
3452377 c.1317C>T p.P439P Chr.11: 674722-674722 on Build GRCh38
5000514 c.1358G>T p.W453L Chr.11: 674681-674681 on Build GRCh38
1203303 c.1415C>T p.T472M Chr.11: 674624-674624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759129663 Chr.11: 674671-674671 on Build GRCh38
rs767471113 Chr.11: 674748-674748 on Build GRCh38
rs751558688 Chr.11: 674690-674690 on Build GRCh38
rs754832756 Chr.11: 674746-674746 on Build GRCh38
rs766654698 Chr.11: 674664-674664 on Build GRCh38
rs762639219 Chr.11: 674732-674732 on Build GRCh38
rs761183640 Chr.11: 674659-674659 on Build GRCh38
rs373166398 Chr.11: 674711-674711 on Build GRCh38
rs149746031 Chr.11: 674721-674721 on Build GRCh38
rs374222357 Chr.11: 674691-674691 on Build GRCh38
rs777850157 Chr.11: 674788-674788 on Build GRCh38
rs961186748 Chr.11: 674652-674652 on Build GRCh38
rs373286290 Chr.11: 674747-674747 on Build GRCh38
rs755902234 Chr.11: 674764-674764 on Build GRCh38
rs747498431 Chr.11: 674720-674720 on Build GRCh38
rs764182783 Chr.11: 674689-674689 on Build GRCh38
rs200163005 Chr.11: 674758-674758 on Build GRCh38
rs757975287 Chr.11: 674687-674687 on Build GRCh38
rs187523180 Chr.11: 674719-674719 on Build GRCh38
rs777190247 Chr.11: 674806-674806 on Build GRCh38
rs745627333 Chr.11: 674700-674700 on Build GRCh38
rs764961241 Chr.11: 674677-674677 on Build GRCh38
rs147839531 Chr.11: 674731-674731 on Build GRCh38
rs866708744 Chr.11: 674759-674759 on Build GRCh38
rs747111818 Chr.11: 674723-674723 on Build GRCh38
rs7125451 Chr.11: 674733-674733 on Build GRCh38
rs201341584 Chr.11: 674657-674657 on Build GRCh38
rs752294703 Chr.11: 674682-674682 on Build GRCh38
rs758576815 Chr.11: 674767-674767 on Build GRCh38
rs148012724 Chr.11: 674765-674765 on Build GRCh38
rs1049200635 Chr.11: 674656-674656 on Build GRCh38
rs35303725 Chr.11: 674638-674638 on Build GRCh38
rs139679730 Chr.11: 674800-674800 on Build GRCh38
rs544444036 Chr.11: 674668-674668 on Build GRCh38
rs376063833 Chr.11: 674715-674715 on Build GRCh38
rs748296992 Chr.11: 674714-674714 on Build GRCh38
rs753858256 Chr.11: 674722-674722 on Build GRCh38
rs1007134273 Chr.11: 674703-674703 on Build GRCh38
rs34094369 Chr.11: 674675-674675 on Build GRCh38
rs1013773368 Chr.11: 674654-674654 on Build GRCh38
rs376238473 Chr.11: 674789-674789 on Build GRCh38
rs368628431 Chr.11: 674639-674639 on Build GRCh38

View Full Product Details