Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 70486116-70486367 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM135A
Entrez Gene ID
57579
Gene Name
family with sequence similarity 135 member A
Gene Aliases
KIAA1411
UniGene
Hs.211700
Gene Chromosome Location
Chr.6: 70413404-70561174 on Build GRCh38
Transcripts
NM_001105531.2 NM_001162529.1 NM_020819.4 XM_005248743.1 XM_005248744.4 XM_005248745.2 XM_005248748.2 XM_005248749.1 XM_005248751.2 XM_011535993.2 XM_011535994.2 XM_011535995.2 XM_011535996.2 XM_011535997.2 XM_017011124.1 XM_017011125.1 XM_017011126.1 XM_017011127.1 XM_017011128.1 XM_017011129.1 XM_017011130.1 XM_017011131.1 XM_017011132.1 XM_017011133.1 XM_017011134.1 XM_017011135.1 XM_017011136.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6658505 c.724C>A p.R242S Chr.6: 70486196-70486196 on Build GRCh38
4987642 c.724C>T p.R242C Chr.6: 70486196-70486196 on Build GRCh38
1081260 c.749G>A p.R250Q Chr.6: 70486221-70486221 on Build GRCh38
5633621 c.736A>G p.R246G Chr.6: 70486208-70486208 on Build GRCh38
1081259 c.730C>A p.L244I Chr.6: 70486202-70486202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766346790 Chr.6: 70486220-70486220 on Build GRCh38
rs183667677 Chr.6: 70486314-70486314 on Build GRCh38
rs779495844 Chr.6: 70486254-70486254 on Build GRCh38
rs769475515 Chr.6: 70486212-70486212 on Build GRCh38
rs180687505 Chr.6: 70486307-70486307 on Build GRCh38
rs747858200 Chr.6: 70486270-70486270 on Build GRCh38
rs746015316 Chr.6: 70486305-70486305 on Build GRCh38
rs565361768 Chr.6: 70486332-70486332 on Build GRCh38
rs369691884 Chr.6: 70486221-70486221 on Build GRCh38
rs377070236 Chr.6: 70486260-70486260 on Build GRCh38
rs767177273 Chr.6: 70486225-70486225 on Build GRCh38
rs770301005 Chr.6: 70486290-70486290 on Build GRCh38
rs780255288 Chr.6: 70486267-70486267 on Build GRCh38
rs1014909945 Chr.6: 70486288-70486288 on Build GRCh38
rs142935920 Chr.6: 70486189-70486189 on Build GRCh38
rs142154928 Chr.6: 70486215-70486215 on Build GRCh38
rs772935543 Chr.6: 70486276-70486276 on Build GRCh38
rs772832673 Chr.6: 70486214-70486214 on Build GRCh38
rs757225680 Chr.6: 70486253-70486253 on Build GRCh38
rs144036955 Chr.6: 70486238-70486238 on Build GRCh38
rs759516112 Chr.6: 70486275-70486275 on Build GRCh38
rs757755738 Chr.6: 70486186-70486186 on Build GRCh38
rs772275248 Chr.6: 70486204-70486204 on Build GRCh38
rs200570801 Chr.6: 70486223-70486223 on Build GRCh38
rs746256854 Chr.6: 70486196-70486196 on Build GRCh38
rs576341362 Chr.6: 70486313-70486313 on Build GRCh38
rs749844146 Chr.6: 70486178-70486178 on Build GRCh38
rs535171868 Chr.6: 70486151-70486151 on Build GRCh38
rs747768113 Chr.6: 70486209-70486209 on Build GRCh38
rs753917224 Chr.6: 70486249-70486249 on Build GRCh38
rs1010135458 Chr.6: 70486211-70486211 on Build GRCh38
rs776373240 Chr.6: 70486205-70486205 on Build GRCh38
rs139238765 Chr.6: 70486230-70486230 on Build GRCh38
rs756217306 Chr.6: 70486157-70486157 on Build GRCh38
rs781495600 Chr.6: 70486148-70486148 on Build GRCh38
rs1051120234 Chr.6: 70486316-70486316 on Build GRCh38
rs375311087 Chr.6: 70486231-70486231 on Build GRCh38
rs764218702 Chr.6: 70486248-70486248 on Build GRCh38
rs752868530 Chr.6: 70486156-70486156 on Build GRCh38
rs748823560 Chr.6: 70486280-70486280 on Build GRCh38
rs972256854 Chr.6: 70486304-70486304 on Build GRCh38
rs769563805 Chr.6: 70486274-70486274 on Build GRCh38
rs756309054 Chr.6: 70486247-70486247 on Build GRCh38
rs777801284 Chr.6: 70486170-70486170 on Build GRCh38

View Full Product Details