Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38395250-38395505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SOCS7
Entrez Gene ID
30837
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
NAP4, NCKAP4
UniGene
Hs.514132
Gene Chromosome Location
Chr.17: 38351876-38405593 on Build GRCh38
Transcripts
NM_014598.3 XM_017024551.1 XM_017024552.1

SNP Detail

SNP ID Chromosome Location
rs138279795 Chr.17: 38395331-38395331 on Build GRCh38
rs754044452 Chr.17: 38395406-38395406 on Build GRCh38
rs904062481 Chr.17: 38395384-38395385 on Build GRCh38
rs780827348 Chr.17: 38395317-38395317 on Build GRCh38
rs762177660 Chr.17: 38395361-38395361 on Build GRCh38
rs781257536 Chr.17: 38395444-38395444 on Build GRCh38
rs773883789 Chr.17: 38395360-38395360 on Build GRCh38
rs904017567 Chr.17: 38395402-38395402 on Build GRCh38
rs564738496 Chr.17: 38395288-38395288 on Build GRCh38
rs761171384 Chr.17: 38395392-38395392 on Build GRCh38
rs750169490 Chr.17: 38395455-38395455 on Build GRCh38
rs201020366 Chr.17: 38395373-38395373 on Build GRCh38
rs768123387 Chr.17: 38395366-38395366 on Build GRCh38
rs748955216 Chr.17: 38395337-38395337 on Build GRCh38
rs200796575 Chr.17: 38395486-38395486 on Build GRCh38
rs541770563 Chr.17: 38395278-38395278 on Build GRCh38
rs61744884 Chr.17: 38395334-38395334 on Build GRCh38
rs929587620 Chr.17: 38395304-38395304 on Build GRCh38
rs755279853 Chr.17: 38395407-38395407 on Build GRCh38
rs768089703 Chr.17: 38395358-38395358 on Build GRCh38
rs769732574 Chr.17: 38395467-38395467 on Build GRCh38
rs999660027 Chr.17: 38395469-38395469 on Build GRCh38
rs201401274 Chr.17: 38395303-38395303 on Build GRCh38
rs1012730856 Chr.17: 38395355-38395355 on Build GRCh38
rs757237424 Chr.17: 38395316-38395316 on Build GRCh38
rs757111366 Chr.17: 38395443-38395443 on Build GRCh38
rs748902008 Chr.17: 38395477-38395477 on Build GRCh38
rs746899966 Chr.17: 38395299-38395299 on Build GRCh38
rs766698602 Chr.17: 38395393-38395393 on Build GRCh38
rs748339292 Chr.17: 38395325-38395325 on Build GRCh38
rs755878975 Chr.17: 38395462-38395462 on Build GRCh38
rs752866284 Chr.17: 38395427-38395427 on Build GRCh38

View Full Product Details