Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 124527114-124527373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTNAP5
Entrez Gene ID
129684
Gene Name
contactin associated protein like 5
Gene Aliases
caspr5
UniGene
Hs.660653
Gene Chromosome Location
Chr.2: 124025287-124921201 on Build GRCh38
Transcripts
NM_130773.3 XM_006712258.1 XM_017003316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3041955 c.1528C>A p.Q510K Chr.2: 124527338-124527338 on Build GRCh38
350363 c.1530A>T p.Q510H Chr.2: 124527340-124527340 on Build GRCh38
3041954 c.1512T>C p.N504N Chr.2: 124527322-124527322 on Build GRCh38
3727850 c.1475-2A>T p.? Chr.2: 124527283-124527283 on Build GRCh38
1243165 c.1482C>T p.P494P Chr.2: 124527292-124527292 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs963185789 Chr.2: 124527279-124527279 on Build GRCh38
rs759836169 Chr.2: 124527249-124527249 on Build GRCh38
rs544788974 Chr.2: 124527152-124527152 on Build GRCh38
rs201841459 Chr.2: 124527304-124527304 on Build GRCh38
rs772926173 Chr.2: 124527331-124527331 on Build GRCh38
rs998885907 Chr.2: 124527184-124527184 on Build GRCh38
rs778085287 Chr.2: 124527298-124527298 on Build GRCh38
rs73956310 Chr.2: 124527237-124527237 on Build GRCh38
rs771053803 Chr.2: 124527300-124527300 on Build GRCh38
rs777689162 Chr.2: 124527239-124527239 on Build GRCh38
rs774841803 Chr.2: 124527246-124527246 on Build GRCh38
rs756532115 Chr.2: 124527264-124527264 on Build GRCh38
rs774075983 Chr.2: 124527340-124527340 on Build GRCh38
rs772411421 Chr.2: 124527252-124527252 on Build GRCh38
rs759519999 Chr.2: 124527346-124527346 on Build GRCh38
rs960193036 Chr.2: 124527196-124527196 on Build GRCh38
rs772608952 Chr.2: 124527305-124527305 on Build GRCh38
rs778110920 Chr.2: 124527289-124527289 on Build GRCh38
rs775078906 Chr.2: 124527165-124527165 on Build GRCh38
rs564765950 Chr.2: 124527203-124527203 on Build GRCh38
rs749556691 Chr.2: 124527292-124527292 on Build GRCh38
rs765978315 Chr.2: 124527337-124527337 on Build GRCh38
rs1032373933 Chr.2: 124527224-124527224 on Build GRCh38
rs376836000 Chr.2: 124527282-124527282 on Build GRCh38
rs761341439 Chr.2: 124527259-124527259 on Build GRCh38
rs749854227 Chr.2: 124527269-124527269 on Build GRCh38
rs938276385 Chr.2: 124527141-124527141 on Build GRCh38
rs769026647 Chr.2: 124527254-124527256 on Build GRCh38
rs752582069 Chr.2: 124527295-124527295 on Build GRCh38
rs539898795 Chr.2: 124527293-124527293 on Build GRCh38
rs760394749 Chr.2: 124527222-124527222 on Build GRCh38
rs781488471 Chr.2: 124527225-124527225 on Build GRCh38
rs762314550 Chr.2: 124527333-124527333 on Build GRCh38
rs775900892 Chr.2: 124527257-124527257 on Build GRCh38
rs991041145 Chr.2: 124527313-124527313 on Build GRCh38
rs751560819 Chr.2: 124527286-124527286 on Build GRCh38
rs776920720 Chr.2: 124527264-124527265 on Build GRCh38
rs749405963 Chr.2: 124527240-124527240 on Build GRCh38
rs149728848 Chr.2: 124527226-124527226 on Build GRCh38
rs371501785 Chr.2: 124527303-124527303 on Build GRCh38
rs902638944 Chr.2: 124527210-124527210 on Build GRCh38
rs926978255 Chr.2: 124527297-124527297 on Build GRCh38
rs963633848 Chr.2: 124527216-124527216 on Build GRCh38
rs764991545 Chr.2: 124527265-124527265 on Build GRCh38
rs1035851553 Chr.2: 124527189-124527189 on Build GRCh38
rs771300195 Chr.2: 124527242-124527242 on Build GRCh38
rs200602463 Chr.2: 124527326-124527326 on Build GRCh38
rs553166656 Chr.2: 124527207-124527207 on Build GRCh38
rs775810877 Chr.2: 124527323-124527323 on Build GRCh38
rs879452040 Chr.2: 124527205-124527205 on Build GRCh38
rs749258395 Chr.2: 124527299-124527299 on Build GRCh38
rs1015070390 Chr.2: 124527275-124527275 on Build GRCh38
rs896648214 Chr.2: 124527158-124527158 on Build GRCh38
rs991576157 Chr.2: 124527201-124527201 on Build GRCh38
rs529698269 Chr.2: 124527258-124527258 on Build GRCh38

View Full Product Details