Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25239059-25239306 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNMT3A
Entrez Gene ID
1788
Gene Name
DNA methyltransferase 3 alpha
Gene Aliases
DNMT3A2, M.HsaIIIA, TBRS
UniGene
Hs.515840
Gene Chromosome Location
Chr.2: 25232961-25342590 on Build GRCh38
Transcripts
NM_001320893.1 NM_022552.4 NM_153759.3 NM_175629.2 NR_135490.1 XM_005264175.4 XM_005264177.4 XM_011532662.2 XM_011532663.2 XM_011532664.2 XM_011532666.2 XM_011532667.2 XM_017003526.1 XM_017003527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6907881 c.2387G>A p.G796D Chr.2: 25239151-25239151 on Build GRCh38
7339083 c.2390A>G p.N797S Chr.2: 25239148-25239148 on Build GRCh38
5735968 c.2371G>A p.A791T Chr.2: 25239167-25239167 on Build GRCh38
6907835 c.2350G>T p.E784* Chr.2: 25239188-25239188 on Build GRCh38
4991716 c.1770G>A p.M590I Chr.2: 25239201-25239201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754227840 Chr.2: 25239256-25239271 on Build GRCh38
rs901395842 Chr.2: 25239136-25239136 on Build GRCh38
rs998597777 Chr.2: 25239141-25239141 on Build GRCh38
rs368403376 Chr.2: 25239271-25239271 on Build GRCh38
rs774190744 Chr.2: 25239104-25239104 on Build GRCh38
rs778123826 Chr.2: 25239232-25239232 on Build GRCh38
rs774100557 Chr.2: 25239163-25239163 on Build GRCh38
rs372962849 Chr.2: 25239101-25239101 on Build GRCh38
rs556021342 Chr.2: 25239268-25239268 on Build GRCh38
rs766960606 Chr.2: 25239115-25239115 on Build GRCh38
rs756261389 Chr.2: 25239231-25239231 on Build GRCh38
rs775989176 Chr.2: 25239208-25239208 on Build GRCh38
rs781138077 Chr.2: 25239151-25239151 on Build GRCh38
rs749729041 Chr.2: 25239102-25239102 on Build GRCh38
rs770526583 Chr.2: 25239157-25239158 on Build GRCh38
rs774168244 Chr.2: 25239177-25239178 on Build GRCh38
rs140581391 Chr.2: 25239270-25239270 on Build GRCh38
rs748836168 Chr.2: 25239148-25239148 on Build GRCh38
rs886148050 Chr.2: 25239141-25239141 on Build GRCh38
rs759097873 Chr.2: 25239106-25239106 on Build GRCh38
rs764146514 Chr.2: 25239130-25239130 on Build GRCh38
rs756566100 Chr.2: 25239154-25239154 on Build GRCh38
rs757083492 Chr.2: 25239140-25239140 on Build GRCh38
rs375845138 Chr.2: 25239179-25239179 on Build GRCh38
rs777515541 Chr.2: 25239107-25239108 on Build GRCh38
rs775429193 Chr.2: 25239116-25239116 on Build GRCh38
rs770706826 Chr.2: 25239103-25239103 on Build GRCh38
rs754420694 Chr.2: 25239233-25239233 on Build GRCh38
rs757658409 Chr.2: 25239259-25239259 on Build GRCh38
rs375129631 Chr.2: 25239266-25239266 on Build GRCh38
rs771474959 Chr.2: 25239158-25239158 on Build GRCh38
rs765045799 Chr.2: 25239217-25239217 on Build GRCh38
rs771615148 Chr.2: 25239267-25239267 on Build GRCh38
rs760932038 Chr.2: 25239217-25239217 on Build GRCh38
rs770131563 Chr.2: 25239097-25239097 on Build GRCh38
rs370751539 Chr.2: 25239199-25239199 on Build GRCh38
rs1025684646 Chr.2: 25239151-25239151 on Build GRCh38
rs759692474 Chr.2: 25239225-25239225 on Build GRCh38
rs74708853 Chr.2: 25239215-25239215 on Build GRCh38
rs778414705 Chr.2: 25239155-25239155 on Build GRCh38
rs759311226 Chr.2: 25239184-25239184 on Build GRCh38
rs764672718 Chr.2: 25239224-25239225 on Build GRCh38
rs745517428 Chr.2: 25239165-25239165 on Build GRCh38
rs774846521 Chr.2: 25239180-25239180 on Build GRCh38
rs755013083 Chr.2: 25239149-25239149 on Build GRCh38
rs760540599 Chr.2: 25239125-25239125 on Build GRCh38
rs758014471 Chr.2: 25239207-25239207 on Build GRCh38
rs766564570 Chr.2: 25239143-25239143 on Build GRCh38
rs771816732 Chr.2: 25239199-25239201 on Build GRCh38
rs962973415 Chr.2: 25239181-25239181 on Build GRCh38
rs1005623505 Chr.2: 25239167-25239167 on Build GRCh38
rs542806749 Chr.2: 25239227-25239227 on Build GRCh38
rs375038430 Chr.2: 25239273-25239273 on Build GRCh38
rs753567076 Chr.2: 25239137-25239137 on Build GRCh38
rs979932565 Chr.2: 25239205-25239205 on Build GRCh38
rs1004794515 Chr.2: 25239142-25239142 on Build GRCh38
rs752626029 Chr.2: 25239208-25239208 on Build GRCh38
rs767588813 Chr.2: 25239148-25239148 on Build GRCh38

View Full Product Details