Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 135009302-135009559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER3
Entrez Gene ID
347732
Gene Name
cation channel sperm associated 3
Gene Aliases
CACRC
UniGene
Hs.631804
Gene Chromosome Location
Chr.5: 134967906-135011707 on Build GRCh38
Transcripts
NM_178019.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1237164 c.918C>T p.S306S Chr.5: 135009472-135009472 on Build GRCh38
735649 c.841G>A p.E281K Chr.5: 135009395-135009395 on Build GRCh38
1060958 c.899G>A p.R300Q Chr.5: 135009453-135009453 on Build GRCh38
6226275 c.874G>A p.G292R Chr.5: 135009428-135009428 on Build GRCh38
6876076 c.892C>T p.L298L Chr.5: 135009446-135009446 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs375830682 Chr.5: 135009399-135009399 on Build GRCh38
rs780658228 Chr.5: 135009477-135009477 on Build GRCh38
rs770334206 Chr.5: 135009382-135009382 on Build GRCh38
rs372274284 Chr.5: 135009356-135009356 on Build GRCh38
rs779928001 Chr.5: 135009493-135009493 on Build GRCh38
rs947013752 Chr.5: 135009538-135009538 on Build GRCh38
rs372723476 Chr.5: 135009395-135009395 on Build GRCh38
rs564473634 Chr.5: 135009368-135009368 on Build GRCh38
rs904215301 Chr.5: 135009374-135009374 on Build GRCh38
rs753699833 Chr.5: 135009363-135009363 on Build GRCh38
rs778760254 Chr.5: 135009367-135009367 on Build GRCh38
rs866126601 Chr.5: 135009344-135009344 on Build GRCh38
rs756527609 Chr.5: 135009476-135009476 on Build GRCh38
rs775674797 Chr.5: 135009385-135009385 on Build GRCh38
rs141786005 Chr.5: 135009373-135009373 on Build GRCh38
rs764327471 Chr.5: 135009331-135009331 on Build GRCh38
rs763091773 Chr.5: 135009324-135009324 on Build GRCh38
rs927170532 Chr.5: 135009505-135009505 on Build GRCh38
rs149771301 Chr.5: 135009469-135009469 on Build GRCh38
rs142712287 Chr.5: 135009452-135009452 on Build GRCh38
rs373841452 Chr.5: 135009503-135009503 on Build GRCh38
rs776774155 Chr.5: 135009531-135009531 on Build GRCh38
rs562430942 Chr.5: 135009393-135009393 on Build GRCh38
rs151000651 Chr.5: 135009453-135009453 on Build GRCh38
rs769069112 Chr.5: 135009483-135009483 on Build GRCh38
rs766198513 Chr.5: 135009522-135009522 on Build GRCh38
rs142494932 Chr.5: 135009435-135009435 on Build GRCh38
rs370006694 Chr.5: 135009446-135009446 on Build GRCh38
rs760861523 Chr.5: 135009508-135009508 on Build GRCh38
rs780977259 Chr.5: 135009512-135009512 on Build GRCh38
rs749650858 Chr.5: 135009478-135009478 on Build GRCh38
rs369037609 Chr.5: 135009392-135009392 on Build GRCh38
rs201904369 Chr.5: 135009534-135009534 on Build GRCh38
rs139694304 Chr.5: 135009466-135009466 on Build GRCh38
rs372927854 Chr.5: 135009423-135009423 on Build GRCh38
rs760412285 Chr.5: 135009459-135009461 on Build GRCh38
rs376864680 Chr.5: 135009357-135009357 on Build GRCh38
rs772864391 Chr.5: 135009380-135009380 on Build GRCh38
rs369957678 Chr.5: 135009499-135009499 on Build GRCh38
rs751586672 Chr.5: 135009332-135009332 on Build GRCh38
rs187422354 Chr.5: 135009535-135009535 on Build GRCh38
rs982295980 Chr.5: 135009461-135009461 on Build GRCh38
rs767442635 Chr.5: 135009403-135009403 on Build GRCh38
rs368336545 Chr.5: 135009341-135009341 on Build GRCh38
rs1011460819 Chr.5: 135009333-135009333 on Build GRCh38
rs761923156 Chr.5: 135009335-135009335 on Build GRCh38
rs893079247 Chr.5: 135009326-135009326 on Build GRCh38
rs146238260 Chr.5: 135009377-135009377 on Build GRCh38
rs72800379 Chr.5: 135009372-135009372 on Build GRCh38
rs775932166 Chr.5: 135009323-135009323 on Build GRCh38
rs773350193 Chr.5: 135009407-135009407 on Build GRCh38

View Full Product Details