Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3639686-3639941 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ART5
Entrez Gene ID
116969
Gene Name
ADP-ribosyltransferase 5
Gene Aliases
ARTC5
UniGene
Hs.125680
Gene Chromosome Location
Chr.11: 3638503-3642392 on Build GRCh38
Transcripts
NM_001079536.1 NM_001297668.1 NM_053017.4 NR_123732.1 XM_005252779.3 XM_006718136.3 XM_011519879.2 XM_017017171.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7100211 c.530G>A p.R177H Chr.11: 3639899-3639899 on Build GRCh38
7400205 c.603T>A p.S201S Chr.11: 3639826-3639826 on Build GRCh38
4860168 c.520G>T p.D174Y Chr.11: 3639909-3639909 on Build GRCh38
7425968 c.688T>A p.F230I Chr.11: 3639741-3639741 on Build GRCh38
3752354 c.672T>C p.I224I Chr.11: 3639757-3639757 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773024564 Chr.11: 3639824-3639827 on Build GRCh38
rs775225474 Chr.11: 3639748-3639748 on Build GRCh38
rs768221945 Chr.11: 3639875-3639875 on Build GRCh38
rs144895632 Chr.11: 3639845-3639845 on Build GRCh38
rs376801553 Chr.11: 3639716-3639716 on Build GRCh38
rs776313535 Chr.11: 3639709-3639709 on Build GRCh38
rs143414771 Chr.11: 3639713-3639713 on Build GRCh38
rs758591505 Chr.11: 3639765-3639765 on Build GRCh38
rs17851204 Chr.11: 3639907-3639907 on Build GRCh38
rs769769262 Chr.11: 3639773-3639773 on Build GRCh38
rs781119812 Chr.11: 3639835-3639835 on Build GRCh38
rs768331757 Chr.11: 3639786-3639786 on Build GRCh38
rs756087101 Chr.11: 3639844-3639844 on Build GRCh38
rs149052085 Chr.11: 3639899-3639899 on Build GRCh38
rs773752380 Chr.11: 3639789-3639789 on Build GRCh38
rs796103332 Chr.11: 3639905-3639905 on Build GRCh38
rs751597729 Chr.11: 3639825-3639825 on Build GRCh38
rs769574376 Chr.11: 3639727-3639727 on Build GRCh38
rs61740976 Chr.11: 3639822-3639822 on Build GRCh38
rs777061748 Chr.11: 3639796-3639796 on Build GRCh38
rs760036205 Chr.11: 3639804-3639804 on Build GRCh38
rs372472425 Chr.11: 3639752-3639752 on Build GRCh38
rs903579591 Chr.11: 3639908-3639908 on Build GRCh38
rs762733263 Chr.11: 3639780-3639780 on Build GRCh38
rs758574280 Chr.11: 3639821-3639821 on Build GRCh38
rs753040292 Chr.11: 3639807-3639807 on Build GRCh38
rs555998435 Chr.11: 3639770-3639770 on Build GRCh38
rs766796058 Chr.11: 3639793-3639793 on Build GRCh38
rs774113971 Chr.11: 3639749-3639750 on Build GRCh38
rs74708481 Chr.11: 3639750-3639750 on Build GRCh38
rs779116836 Chr.11: 3639760-3639760 on Build GRCh38
rs146749287 Chr.11: 3639758-3639758 on Build GRCh38
rs752987241 Chr.11: 3639763-3639763 on Build GRCh38
rs772682500 Chr.11: 3639900-3639900 on Build GRCh38
rs143991969 Chr.11: 3639775-3639775 on Build GRCh38
rs773985453 Chr.11: 3639884-3639884 on Build GRCh38
rs748137393 Chr.11: 3639750-3639750 on Build GRCh38
rs143829651 Chr.11: 3639767-3639768 on Build GRCh38
rs929551924 Chr.11: 3639829-3639829 on Build GRCh38
rs751728239 Chr.11: 3639834-3639834 on Build GRCh38
rs200329371 Chr.11: 3639806-3639806 on Build GRCh38
rs770807658 Chr.11: 3639769-3639769 on Build GRCh38
rs745866899 Chr.11: 3639841-3639841 on Build GRCh38
rs937888264 Chr.11: 3639823-3639823 on Build GRCh38
rs777767064 Chr.11: 3639766-3639766 on Build GRCh38
rs747621214 Chr.11: 3639885-3639885 on Build GRCh38
rs866613845 Chr.11: 3639851-3639851 on Build GRCh38
rs754230688 Chr.11: 3639756-3639756 on Build GRCh38
rs749042074 Chr.11: 3639854-3639854 on Build GRCh38
rs140453940 Chr.11: 3639790-3639790 on Build GRCh38
rs763578150 Chr.11: 3639751-3639751 on Build GRCh38
rs943545943 Chr.11: 3639794-3639794 on Build GRCh38
rs202122052 Chr.11: 3639768-3639768 on Build GRCh38
rs763312413 Chr.11: 3639912-3639912 on Build GRCh38
rs4910876 Chr.11: 3639910-3639910 on Build GRCh38
rs143088148 Chr.11: 3639909-3639909 on Build GRCh38
rs764236648 Chr.11: 3639916-3639916 on Build GRCh38

View Full Product Details