Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 7992800-7993073 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3F
Entrez Gene ID
8665
Gene Name
eukaryotic translation initiation factor 3 subunit F
Gene Aliases
EIF3S5, eIF3-p47
UniGene
Hs.516023
Gene Chromosome Location
Chr.11: 7987320-7996171 on Build GRCh38
Transcripts
NM_003754.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3870301 c.627C>T p.A209A Chr.11: 7992953-7992953 on Build GRCh38
327354 c.549_550delTG p.V184fs*61 Chr.11: 7992920-7992921 on Build GRCh38
3870299 c.568A>C p.T190P Chr.11: 7992894-7992894 on Build GRCh38
6364706 c.547T>C p.S183P Chr.11: 7992918-7992918 on Build GRCh38
5680442 c.636C>G p.S212R Chr.11: 7993007-7993007 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906340250 Chr.11: 7992999-7992999 on Build GRCh38
rs765901651 Chr.11: 7993052-7993052 on Build GRCh38
rs371389618 Chr.11: 7993000-7993000 on Build GRCh38
rs189875301 Chr.11: 7993020-7993020 on Build GRCh38
rs761446402 Chr.11: 7992954-7992954 on Build GRCh38
rs751012517 Chr.11: 7992868-7992870 on Build GRCh38
rs777683450 Chr.11: 7992890-7992890 on Build GRCh38
rs987401450 Chr.11: 7992894-7992894 on Build GRCh38
rs781150083 Chr.11: 7992876-7992876 on Build GRCh38
rs376154162 Chr.11: 7992895-7992895 on Build GRCh38
rs747423679 Chr.11: 7993040-7993040 on Build GRCh38
rs781131467 Chr.11: 7993004-7993004 on Build GRCh38
rs749438716 Chr.11: 7992893-7992893 on Build GRCh38
rs552001240 Chr.11: 7993042-7993042 on Build GRCh38
rs146896448 Chr.11: 7992821-7992821 on Build GRCh38
rs780085811 Chr.11: 7992861-7992861 on Build GRCh38
rs751470741 Chr.11: 7992873-7992873 on Build GRCh38
rs753419211 Chr.11: 7992836-7992836 on Build GRCh38
rs143747261 Chr.11: 7992962-7992962 on Build GRCh38
rs147597507 Chr.11: 7992896-7992896 on Build GRCh38
rs761466751 Chr.11: 7992839-7992839 on Build GRCh38
rs746357618 Chr.11: 7993032-7993032 on Build GRCh38
rs762474947 Chr.11: 7992974-7992974 on Build GRCh38
rs754778091 Chr.11: 7992875-7992875 on Build GRCh38
rs950500853 Chr.11: 7992891-7992891 on Build GRCh38
rs748369599 Chr.11: 7992882-7992882 on Build GRCh38
rs750015811 Chr.11: 7992844-7992844 on Build GRCh38
rs756387297 Chr.11: 7993008-7993008 on Build GRCh38
rs1057510981 Chr.11: 7993018-7993018 on Build GRCh38
rs148143652 Chr.11: 7993019-7993019 on Build GRCh38
rs764713018 Chr.11: 7992843-7992843 on Build GRCh38
rs772880147 Chr.11: 7993050-7993050 on Build GRCh38
rs200275224 Chr.11: 7993027-7993027 on Build GRCh38
rs765520142 Chr.11: 7993039-7993039 on Build GRCh38
rs758233181 Chr.11: 7992851-7992851 on Build GRCh38
rs775886835 Chr.11: 7992950-7992950 on Build GRCh38
rs760042710 Chr.11: 7992901-7992901 on Build GRCh38
rs765836648 Chr.11: 7992975-7992975 on Build GRCh38
rs770209672 Chr.11: 7992884-7992884 on Build GRCh38
rs752580818 Chr.11: 7993007-7993007 on Build GRCh38
rs1042086841 Chr.11: 7992996-7992996 on Build GRCh38
rs749877936 Chr.11: 7992963-7992963 on Build GRCh38
rs994933186 Chr.11: 7993044-7993044 on Build GRCh38
rs912844816 Chr.11: 7992830-7992830 on Build GRCh38
rs201463294 Chr.11: 7992976-7992976 on Build GRCh38
rs896832401 Chr.11: 7992921-7992921 on Build GRCh38
rs751457916 Chr.11: 7992995-7992995 on Build GRCh38
rs762514866 Chr.11: 7993051-7993051 on Build GRCh38
rs4758267 Chr.11: 7992932-7992932 on Build GRCh38
rs943057377 Chr.11: 7992929-7992929 on Build GRCh38

View Full Product Details