Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18960459-18960708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC5A10
Entrez Gene ID
125206
Gene Name
solute carrier family 5 member 10
Gene Aliases
SGLT-5, SGLT5
UniGene
Hs.462418
Gene Chromosome Location
Chr.17: 18950676-19020691 on Build GRCh38
Transcripts
NM_001042450.2 NM_001270648.1 NM_001270649.1 NM_001282417.1 NM_152351.4 XM_011523652.1 XM_011523653.2 XM_017024191.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6783037 c.453G>A p.S151S Chr.17: 18960652-18960652 on Build GRCh38
5851989 c.360delA p.L120fs*73 Chr.17: 18960559-18960559 on Build GRCh38
6855176 c.183C>T p.I61I Chr.17: 18960550-18960550 on Build GRCh38
6783029 c.393G>T p.Q131H Chr.17: 18960592-18960592 on Build GRCh38
341090 c.362C>A p.P121H Chr.17: 18960561-18960561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs565320412 Chr.17: 18960540-18960540 on Build GRCh38
rs267604770 Chr.17: 18960551-18960551 on Build GRCh38
rs771664719 Chr.17: 18960665-18960665 on Build GRCh38
rs771445304 Chr.17: 18960580-18960580 on Build GRCh38
rs766605517 Chr.17: 18960605-18960605 on Build GRCh38
rs536465559 Chr.17: 18960652-18960652 on Build GRCh38
rs767441512 Chr.17: 18960629-18960632 on Build GRCh38
rs547915569 Chr.17: 18960600-18960600 on Build GRCh38
rs940701102 Chr.17: 18960603-18960603 on Build GRCh38
rs770712809 Chr.17: 18960573-18960573 on Build GRCh38
rs1005723053 Chr.17: 18960632-18960649 on Build GRCh38
rs777696794 Chr.17: 18960567-18960567 on Build GRCh38
rs777239029 Chr.17: 18960584-18960584 on Build GRCh38
rs138879652 Chr.17: 18960568-18960568 on Build GRCh38
rs748003428 Chr.17: 18960556-18960556 on Build GRCh38
rs781032922 Chr.17: 18960681-18960681 on Build GRCh38
rs374489519 Chr.17: 18960509-18960509 on Build GRCh38
rs774682506 Chr.17: 18960688-18960688 on Build GRCh38
rs763336459 Chr.17: 18960668-18960668 on Build GRCh38
rs963747931 Chr.17: 18960548-18960548 on Build GRCh38
rs978080467 Chr.17: 18960650-18960650 on Build GRCh38
rs759698165 Chr.17: 18960625-18960625 on Build GRCh38
rs2074274 Chr.17: 18960666-18960666 on Build GRCh38
rs142113789 Chr.17: 18960595-18960595 on Build GRCh38
rs756115651 Chr.17: 18960649-18960649 on Build GRCh38
rs185855888 Chr.17: 18960578-18960578 on Build GRCh38
rs754960897 Chr.17: 18960550-18960550 on Build GRCh38
rs139579009 Chr.17: 18960651-18960651 on Build GRCh38
rs773565198 Chr.17: 18960599-18960599 on Build GRCh38
rs377718527 Chr.17: 18960616-18960616 on Build GRCh38
rs751689685 Chr.17: 18960612-18960612 on Build GRCh38
rs1022031472 Chr.17: 18960539-18960539 on Build GRCh38
rs948130711 Chr.17: 18960631-18960631 on Build GRCh38
rs200545519 Chr.17: 18960525-18960525 on Build GRCh38
rs756020471 Chr.17: 18960565-18960565 on Build GRCh38
rs116620981 Chr.17: 18960640-18960640 on Build GRCh38
rs767471440 Chr.17: 18960638-18960638 on Build GRCh38
rs142768495 Chr.17: 18960654-18960654 on Build GRCh38
rs1010522259 Chr.17: 18960490-18960490 on Build GRCh38
rs745480583 Chr.17: 18960662-18960662 on Build GRCh38
rs771165564 Chr.17: 18960684-18960684 on Build GRCh38
rs773817698 Chr.17: 18960579-18960579 on Build GRCh38
rs899564160 Chr.17: 18960594-18960594 on Build GRCh38
rs750698199 Chr.17: 18960520-18960520 on Build GRCh38
rs751668877 Chr.17: 18960543-18960543 on Build GRCh38
rs773656939 Chr.17: 18960667-18960667 on Build GRCh38
rs778660053 Chr.17: 18960660-18960660 on Build GRCh38
rs759527570 Chr.17: 18960579-18960586 on Build GRCh38

View Full Product Details