Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 29338083-29338354 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP4
Entrez Gene ID
1846
Gene Name
dual specificity phosphatase 4
Gene Aliases
HVH2, MKP-2, MKP2, TYP
UniGene
Hs.417962
Gene Chromosome Location
Chr.8: 29333062-29350750 on Build GRCh38
Transcripts
NM_001394.6 NM_057158.3 XM_011544428.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2789112 c.521A>G p.Y174C Chr.8: 29338287-29338287 on Build GRCh38
5608308 c.777C>T p.F259F Chr.8: 29338304-29338304 on Build GRCh38
2789115 c.752A>C p.H251P Chr.8: 29338329-29338329 on Build GRCh38
1292671 c.732C>T p.C244C Chr.8: 29338349-29338349 on Build GRCh38
2789111 c.794A>G p.Y265C Chr.8: 29338287-29338287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1001507715 Chr.8: 29338153-29338153 on Build GRCh38
rs772020412 Chr.8: 29338260-29338260 on Build GRCh38
rs369386350 Chr.8: 29338242-29338242 on Build GRCh38
rs768212045 Chr.8: 29338289-29338289 on Build GRCh38
rs1034359451 Chr.8: 29338183-29338183 on Build GRCh38
rs527360327 Chr.8: 29338243-29338243 on Build GRCh38
rs372064347 Chr.8: 29338282-29338282 on Build GRCh38
rs577399149 Chr.8: 29338248-29338248 on Build GRCh38
rs992953391 Chr.8: 29338239-29338239 on Build GRCh38
rs1023455132 Chr.8: 29338219-29338219 on Build GRCh38
rs376576672 Chr.8: 29338283-29338283 on Build GRCh38
rs969650848 Chr.8: 29338226-29338226 on Build GRCh38
rs369227955 Chr.8: 29338303-29338303 on Build GRCh38
rs764888128 Chr.8: 29338321-29338321 on Build GRCh38
rs748514096 Chr.8: 29338314-29338314 on Build GRCh38
rs957961929 Chr.8: 29338155-29338155 on Build GRCh38
rs989692254 Chr.8: 29338157-29338157 on Build GRCh38
rs969061245 Chr.8: 29338150-29338150 on Build GRCh38
rs112844386 Chr.8: 29338253-29338253 on Build GRCh38
rs1021568331 Chr.8: 29338125-29338125 on Build GRCh38
rs199987827 Chr.8: 29338240-29338240 on Build GRCh38

View Full Product Details