Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43342907-43343160 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAL
Entrez Gene ID
161823
Gene Name
adenosine deaminase like
Gene Aliases
-
UniGene
Hs.533913
Gene Chromosome Location
Chr.15: 43330351-43354555 on Build GRCh38
Transcripts
NM_001012969.3 NM_001159280.2 NM_001324364.1 NM_001324365.1 NM_001324366.1 NM_001324367.1 NM_001324368.1 XM_011521291.2 XM_017021964.1 XM_017021965.1 XM_017021966.1 XM_017021967.1 XM_017021968.1 XM_017021969.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6564871 c.409T>G p.L137V Chr.15: 43343059-43343059 on Build GRCh38
1587858 c.355A>C p.K119Q Chr.15: 43343005-43343005 on Build GRCh38
1678406 c.376C>T p.L126F Chr.15: 43343026-43343026 on Build GRCh38
3706583 c.400C>G p.Q134E Chr.15: 43343050-43343050 on Build GRCh38
961944 c.355A>C p.K119Q Chr.15: 43343005-43343005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145228643 Chr.15: 43343002-43343002 on Build GRCh38
rs931906903 Chr.15: 43343033-43343033 on Build GRCh38
rs774758513 Chr.15: 43343017-43343017 on Build GRCh38
rs747827820 Chr.15: 43343103-43343103 on Build GRCh38
rs775694530 Chr.15: 43343041-43343041 on Build GRCh38
rs745856038 Chr.15: 43342950-43342951 on Build GRCh38
rs967070568 Chr.15: 43343001-43343001 on Build GRCh38
rs754661553 Chr.15: 43342996-43342996 on Build GRCh38
rs777310728 Chr.15: 43343114-43343114 on Build GRCh38
rs766792477 Chr.15: 43342957-43342957 on Build GRCh38
rs773770842 Chr.15: 43342945-43342945 on Build GRCh38
rs748802421 Chr.15: 43343115-43343115 on Build GRCh38
rs781055872 Chr.15: 43342998-43342998 on Build GRCh38
rs767723614 Chr.15: 43343040-43343040 on Build GRCh38
rs377641385 Chr.15: 43342972-43342972 on Build GRCh38
rs751391383 Chr.15: 43343087-43343087 on Build GRCh38
rs750395960 Chr.15: 43342971-43342971 on Build GRCh38
rs370771114 Chr.15: 43343013-43343013 on Build GRCh38
rs764264993 Chr.15: 43343050-43343050 on Build GRCh38
rs768610619 Chr.15: 43343064-43343064 on Build GRCh38
rs774398251 Chr.15: 43342958-43342958 on Build GRCh38
rs8041132 Chr.15: 43342938-43342938 on Build GRCh38
rs111429563 Chr.15: 43342978-43342979 on Build GRCh38
rs766282216 Chr.15: 43343085-43343085 on Build GRCh38
rs773547579 Chr.15: 43343004-43343004 on Build GRCh38
rs577489361 Chr.15: 43342986-43342986 on Build GRCh38
rs763173365 Chr.15: 43342952-43342952 on Build GRCh38
rs1050296496 Chr.15: 43343036-43343036 on Build GRCh38
rs990348424 Chr.15: 43342984-43342984 on Build GRCh38
rs747591001 Chr.15: 43342999-43342999 on Build GRCh38
rs373551232 Chr.15: 43343111-43343111 on Build GRCh38
rs370422648 Chr.15: 43343090-43343090 on Build GRCh38
rs200070296 Chr.15: 43343083-43343083 on Build GRCh38
rs554309107 Chr.15: 43342960-43342960 on Build GRCh38
rs11634407 Chr.15: 43343078-43343078 on Build GRCh38
rs201617869 Chr.15: 43342968-43342968 on Build GRCh38
rs749836531 Chr.15: 43343005-43343005 on Build GRCh38
rs746848888 Chr.15: 43342993-43342993 on Build GRCh38
rs759952821 Chr.15: 43343025-43343025 on Build GRCh38
rs149144047 Chr.15: 43343049-43343049 on Build GRCh38

View Full Product Details