Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57610537-57610810 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF25
Entrez Gene ID
115557
Gene Name
Rho guanine nucleotide exchange factor 25
Gene Aliases
GEFT, p63RhoGEF
UniGene
Hs.61581
Gene Chromosome Location
Chr.12: 57610180-57617245 on Build GRCh38
Transcripts
NM_001111270.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5762098 c.151C>T p.R51C Chr.12: 57610621-57610621 on Build GRCh38
6604175 c.180C>T p.R60R Chr.12: 57610650-57610650 on Build GRCh38
468719 c.187G>T p.E63* Chr.12: 57610657-57610657 on Build GRCh38
4795996 c.135C>T p.T45T Chr.12: 57610605-57610605 on Build GRCh38
6859944 c.198C>G p.A66A Chr.12: 57610668-57610668 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750869579 Chr.12: 57610588-57610588 on Build GRCh38
rs764009524 Chr.12: 57610570-57610570 on Build GRCh38
rs760873837 Chr.12: 57610567-57610567 on Build GRCh38
rs776618322 Chr.12: 57610676-57610676 on Build GRCh38
rs756745934 Chr.12: 57610734-57610734 on Build GRCh38
rs368226104 Chr.12: 57610656-57610656 on Build GRCh38
rs775389987 Chr.12: 57610566-57610566 on Build GRCh38
rs750595135 Chr.12: 57610715-57610715 on Build GRCh38
rs771847534 Chr.12: 57610665-57610665 on Build GRCh38
rs773567553 Chr.12: 57610697-57610697 on Build GRCh38
rs765615148 Chr.12: 57610688-57610688 on Build GRCh38
rs763416213 Chr.12: 57610698-57610698 on Build GRCh38
rs953699011 Chr.12: 57610572-57610572 on Build GRCh38
rs751945850 Chr.12: 57610605-57610605 on Build GRCh38
rs761935620 Chr.12: 57610683-57610683 on Build GRCh38
rs145261291 Chr.12: 57610717-57610717 on Build GRCh38
rs746860557 Chr.12: 57610668-57610668 on Build GRCh38
rs909235477 Chr.12: 57610704-57610704 on Build GRCh38
rs201793908 Chr.12: 57610579-57610579 on Build GRCh38
rs756942515 Chr.12: 57610645-57610645 on Build GRCh38
rs201016615 Chr.12: 57610621-57610621 on Build GRCh38
rs1052931007 Chr.12: 57610756-57610756 on Build GRCh38
rs752130294 Chr.12: 57610708-57610708 on Build GRCh38
rs937437198 Chr.12: 57610773-57610773 on Build GRCh38
rs755663287 Chr.12: 57610609-57610609 on Build GRCh38
rs375479122 Chr.12: 57610727-57610727 on Build GRCh38
rs188074637 Chr.12: 57610603-57610603 on Build GRCh38
rs758826647 Chr.12: 57610593-57610593 on Build GRCh38
rs765530080 Chr.12: 57610586-57610586 on Build GRCh38
rs907251704 Chr.12: 57610737-57610737 on Build GRCh38
rs929552263 Chr.12: 57610669-57610669 on Build GRCh38
rs891718824 Chr.12: 57610772-57610772 on Build GRCh38
rs1049544254 Chr.12: 57610672-57610672 on Build GRCh38
rs755398260 Chr.12: 57610715-57610715 on Build GRCh38
rs557599605 Chr.12: 57610751-57610751 on Build GRCh38
rs768800067 Chr.12: 57610675-57610675 on Build GRCh38
rs766792717 Chr.12: 57610699-57610699 on Build GRCh38
rs953170559 Chr.12: 57610603-57610605 on Build GRCh38
rs775408309 Chr.12: 57610666-57610666 on Build GRCh38
rs1045804254 Chr.12: 57610753-57610753 on Build GRCh38
rs748999940 Chr.12: 57610629-57610629 on Build GRCh38
rs7314872 Chr.12: 57610780-57610780 on Build GRCh38
rs776922320 Chr.12: 57610576-57610576 on Build GRCh38

View Full Product Details