Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70394763-70395026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST3GAL2
Entrez Gene ID
6483
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
Gene Aliases
Gal-NAc6S, SIAT4B, ST3GALII, ST3GalA.2
UniGene
Hs.368611
Gene Chromosome Location
Chr.16: 70379435-70439088 on Build GRCh38
Transcripts
NM_006927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
392546 c.528_530delCAT p.I176delI Chr.16: 70394985-70394987 on Build GRCh38
1479050 c.492T>C p.S164S Chr.16: 70395023-70395023 on Build GRCh38
6791058 c.513C>T p.D171D Chr.16: 70395002-70395002 on Build GRCh38
6791057 c.507C>T p.D169D Chr.16: 70395008-70395008 on Build GRCh38
3511369 c.495C>G p.G165G Chr.16: 70395020-70395020 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755393198 Chr.16: 70394938-70394938 on Build GRCh38
rs756839850 Chr.16: 70394965-70394965 on Build GRCh38
rs1051264807 Chr.16: 70394887-70394887 on Build GRCh38
rs923349595 Chr.16: 70394927-70394933 on Build GRCh38
rs746592379 Chr.16: 70394962-70394962 on Build GRCh38
rs749227121 Chr.16: 70394996-70394996 on Build GRCh38
rs534458430 Chr.16: 70394788-70394788 on Build GRCh38
rs890797375 Chr.16: 70394896-70394896 on Build GRCh38
rs374633767 Chr.16: 70394937-70394937 on Build GRCh38
rs940676263 Chr.16: 70394886-70394886 on Build GRCh38
rs918359634 Chr.16: 70394868-70394868 on Build GRCh38
rs939293925 Chr.16: 70394794-70394794 on Build GRCh38
rs766661945 Chr.16: 70395006-70395006 on Build GRCh38
rs753672179 Chr.16: 70394973-70394973 on Build GRCh38
rs577591978 Chr.16: 70394817-70394817 on Build GRCh38
rs937297711 Chr.16: 70394862-70394862 on Build GRCh38
rs146471525 Chr.16: 70395002-70395002 on Build GRCh38
rs777256001 Chr.16: 70394959-70394959 on Build GRCh38
rs199733390 Chr.16: 70394969-70394969 on Build GRCh38
rs555225725 Chr.16: 70394785-70394785 on Build GRCh38
rs776944832 Chr.16: 70395008-70395008 on Build GRCh38
rs376646546 Chr.16: 70394975-70394975 on Build GRCh38
rs984664505 Chr.16: 70394882-70394882 on Build GRCh38
rs372103798 Chr.16: 70394978-70394978 on Build GRCh38
rs8046296 Chr.16: 70394802-70394802 on Build GRCh38
rs371618907 Chr.16: 70394960-70394960 on Build GRCh38
rs753217630 Chr.16: 70394954-70394954 on Build GRCh38
rs768772233 Chr.16: 70394999-70394999 on Build GRCh38
rs567448819 Chr.16: 70394786-70394786 on Build GRCh38
rs766772524 Chr.16: 70394935-70394935 on Build GRCh38
rs981269920 Chr.16: 70394833-70394833 on Build GRCh38
rs199518613 Chr.16: 70394945-70394945 on Build GRCh38
rs148559292 Chr.16: 70394859-70394859 on Build GRCh38
rs141968086 Chr.16: 70394881-70394881 on Build GRCh38
rs540400494 Chr.16: 70395001-70395001 on Build GRCh38
rs547493898 Chr.16: 70394913-70394913 on Build GRCh38
rs759044935 Chr.16: 70394806-70394806 on Build GRCh38
rs941211686 Chr.16: 70394820-70394820 on Build GRCh38
rs542911505 Chr.16: 70394929-70394929 on Build GRCh38
rs575491364 Chr.16: 70394882-70394882 on Build GRCh38
rs376369543 Chr.16: 70394979-70394979 on Build GRCh38

View Full Product Details