Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111306447-111306700 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLCA2
Entrez Gene ID
120376
Gene Name
colorectal cancer associated 2
Gene Aliases
C11orf93, CASC13, LOH11CR1G
UniGene
Hs.31409
Gene Chromosome Location
Chr.11: 111298517-111308735 on Build GRCh38
Transcripts
NM_001136105.2 NM_001271457.1 NM_001271458.1 XM_017017196.1 XM_017017197.1 XM_017017198.1 XM_017017199.1 XM_017017200.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1196186 c.22A>G p.N8D Chr.11: 111306549-111306549 on Build GRCh38
230239 c.7C>T p.P3S Chr.11: 111306534-111306534 on Build GRCh38
4817034 c.63C>T p.F21F Chr.11: 111306590-111306590 on Build GRCh38
2111497 c.50T>G p.F17C Chr.11: 111306577-111306577 on Build GRCh38
4633238 c.52C>A p.P18T Chr.11: 111306579-111306579 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs975423883 Chr.11: 111306594-111306594 on Build GRCh38
rs201037042 Chr.11: 111306577-111306577 on Build GRCh38
rs765129133 Chr.11: 111306649-111306649 on Build GRCh38
rs977309768 Chr.11: 111306666-111306666 on Build GRCh38
rs958514116 Chr.11: 111306599-111306599 on Build GRCh38
rs770553908 Chr.11: 111306495-111306495 on Build GRCh38
rs191344104 Chr.11: 111306483-111306483 on Build GRCh38
rs7109595 Chr.11: 111306494-111306494 on Build GRCh38
rs991310974 Chr.11: 111306630-111306630 on Build GRCh38
rs774053196 Chr.11: 111306506-111306506 on Build GRCh38
rs752812113 Chr.11: 111306527-111306527 on Build GRCh38
rs966412547 Chr.11: 111306507-111306507 on Build GRCh38
rs991817802 Chr.11: 111306503-111306503 on Build GRCh38
rs753674838 Chr.11: 111306641-111306641 on Build GRCh38
rs771566500 Chr.11: 111306540-111306540 on Build GRCh38
rs949948654 Chr.11: 111306650-111306650 on Build GRCh38
rs760633526 Chr.11: 111306628-111306628 on Build GRCh38
rs978292978 Chr.11: 111306543-111306543 on Build GRCh38
rs368278144 Chr.11: 111306659-111306659 on Build GRCh38
rs763989118 Chr.11: 111306636-111306636 on Build GRCh38
rs761767388 Chr.11: 111306647-111306647 on Build GRCh38
rs984888224 Chr.11: 111306562-111306562 on Build GRCh38
rs952150543 Chr.11: 111306531-111306531 on Build GRCh38
rs759189963 Chr.11: 111306536-111306536 on Build GRCh38
rs925414992 Chr.11: 111306557-111306557 on Build GRCh38

View Full Product Details