Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 13364977-13365216 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GFOD1
Entrez Gene ID
54438
Gene Name
glucose-fructose oxidoreductase domain containing 1
Gene Aliases
ADG-90, C6orf114
UniGene
Hs.484686
Gene Chromosome Location
Chr.6: 13357830-13487637 on Build GRCh38
Transcripts
NM_001242628.1 NM_001242630.1 NM_018988.3 XM_011514699.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4912358 c.746T>C p.V249A Chr.6: 13365170-13365170 on Build GRCh38
6054908 c.863C>T p.S288F Chr.6: 13365053-13365053 on Build GRCh38
3697509 c.918C>A p.T306T Chr.6: 13364998-13364998 on Build GRCh38
5032800 c.848C>T p.T283M Chr.6: 13365068-13365068 on Build GRCh38
6405368 c.825G>T p.Q275H Chr.6: 13365091-13365091 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974380914 Chr.6: 13365016-13365016 on Build GRCh38
rs771997138 Chr.6: 13365115-13365115 on Build GRCh38
rs930336687 Chr.6: 13365047-13365047 on Build GRCh38
rs376773882 Chr.6: 13365186-13365186 on Build GRCh38
rs200698442 Chr.6: 13364999-13364999 on Build GRCh38
rs145232878 Chr.6: 13365053-13365053 on Build GRCh38
rs3800487 Chr.6: 13365118-13365118 on Build GRCh38
rs376549071 Chr.6: 13365072-13365072 on Build GRCh38
rs758980110 Chr.6: 13365163-13365163 on Build GRCh38
rs755452446 Chr.6: 13365044-13365044 on Build GRCh38
rs554597063 Chr.6: 13365190-13365190 on Build GRCh38
rs970059313 Chr.6: 13365003-13365003 on Build GRCh38
rs765754263 Chr.6: 13365147-13365147 on Build GRCh38
rs766689068 Chr.6: 13365030-13365030 on Build GRCh38
rs766599346 Chr.6: 13365131-13365131 on Build GRCh38
rs768632234 Chr.6: 13365103-13365103 on Build GRCh38
rs749970186 Chr.6: 13365174-13365174 on Build GRCh38
rs763168816 Chr.6: 13365068-13365068 on Build GRCh38
rs747995227 Chr.6: 13365195-13365195 on Build GRCh38
rs754200124 Chr.6: 13365136-13365136 on Build GRCh38
rs750688356 Chr.6: 13365010-13365010 on Build GRCh38
rs767814561 Chr.6: 13365113-13365113 on Build GRCh38
rs749076810 Chr.6: 13365189-13365189 on Build GRCh38
rs371884277 Chr.6: 13365117-13365117 on Build GRCh38
rs931793694 Chr.6: 13365151-13365151 on Build GRCh38
rs780786102 Chr.6: 13365167-13365167 on Build GRCh38
rs149994474 Chr.6: 13365187-13365187 on Build GRCh38
rs754204541 Chr.6: 13365037-13365037 on Build GRCh38
rs767747080 Chr.6: 13365005-13365005 on Build GRCh38
rs779356979 Chr.6: 13365045-13365045 on Build GRCh38
rs140445771 Chr.6: 13365064-13365064 on Build GRCh38
rs552590164 Chr.6: 13365098-13365098 on Build GRCh38
rs534804617 Chr.6: 13365157-13365157 on Build GRCh38
rs779939486 Chr.6: 13365073-13365073 on Build GRCh38
rs550515114 Chr.6: 13365004-13365004 on Build GRCh38
rs866513622 Chr.6: 13365126-13365126 on Build GRCh38
rs895950029 Chr.6: 13365100-13365100 on Build GRCh38
rs773217142 Chr.6: 13365196-13365196 on Build GRCh38
rs774832011 Chr.6: 13365017-13365017 on Build GRCh38
rs1014843297 Chr.6: 13365127-13365127 on Build GRCh38
rs751050919 Chr.6: 13365051-13365051 on Build GRCh38
rs774191779 Chr.6: 13365107-13365107 on Build GRCh38
rs368584392 Chr.6: 13365193-13365193 on Build GRCh38
rs759860102 Chr.6: 13365146-13365146 on Build GRCh38
rs780776864 Chr.6: 13365058-13365058 on Build GRCh38

View Full Product Details