Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 42934396-42934667 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY3
Entrez Gene ID
10695
Gene Name
canopy FGF signaling regulator 3
Gene Aliases
CAG4A, ERDA5, PRAT4A, TNRC5
UniGene
Hs.414099
Gene Chromosome Location
Chr.6: 42928002-42939287 on Build GRCh38
Transcripts
NM_001318842.1 NM_001318845.1 NM_001318847.1 NM_001318848.1 NM_006586.4 NR_134880.1 NR_134881.1 NR_134882.1 NR_134885.1 NR_134886.1 NR_134888.1 NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1

Gene Symbol
CNPY3-GNMT
Entrez Gene ID
107080644
Gene Name
CNPY3-GNMT readthrough
Gene Aliases
-
UniGene
Hs.144914
Gene Chromosome Location
Chr.6: 42929130-42963880 on Build GRCh38
Transcripts
NM_001318842.1 NM_001318845.1 NM_001318847.1 NM_001318848.1 NM_006586.4 NR_134880.1 NR_134881.1 NR_134882.1 NR_134885.1 NR_134886.1 NR_134888.1 NM_001318856.1 NM_001318857.1 NM_001318858.1 NR_134890.1 NR_134891.1 NR_134892.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6275283 c.169G>T p.V57L Chr.6: 42934492-42934492 on Build GRCh38
5982421 c.177G>A p.L59L Chr.6: 42934500-42934500 on Build GRCh38
6402029 c.194A>G p.E65G Chr.6: 42934517-42934517 on Build GRCh38
1201632 c.199G>A p.G67S Chr.6: 42934522-42934522 on Build GRCh38
1079372 c.224C>T p.T75M Chr.6: 42934547-42934547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146093552 Chr.6: 42934485-42934485 on Build GRCh38
rs760063356 Chr.6: 42934550-42934550 on Build GRCh38
rs754370632 Chr.6: 42934434-42934434 on Build GRCh38
rs556172653 Chr.6: 42934598-42934598 on Build GRCh38
rs199645339 Chr.6: 42934471-42934471 on Build GRCh38
rs753075646 Chr.6: 42934560-42934560 on Build GRCh38
rs765707879 Chr.6: 42934554-42934554 on Build GRCh38
rs750518549 Chr.6: 42934573-42934573 on Build GRCh38
rs769143809 Chr.6: 42934619-42934620 on Build GRCh38
rs753049259 Chr.6: 42934446-42934446 on Build GRCh38
rs376519254 Chr.6: 42934472-42934472 on Build GRCh38
rs761244184 Chr.6: 42934542-42934542 on Build GRCh38
rs375546787 Chr.6: 42934522-42934522 on Build GRCh38
rs766736658 Chr.6: 42934427-42934427 on Build GRCh38
rs979040436 Chr.6: 42934647-42934647 on Build GRCh38
rs1050096392 Chr.6: 42934570-42934570 on Build GRCh38
rs778742820 Chr.6: 42934612-42934612 on Build GRCh38
rs749111912 Chr.6: 42934521-42934521 on Build GRCh38
rs779877202 Chr.6: 42934597-42934597 on Build GRCh38
rs770291121 Chr.6: 42934641-42934641 on Build GRCh38
rs747783493 Chr.6: 42934620-42934620 on Build GRCh38
rs745815310 Chr.6: 42934474-42934474 on Build GRCh38
rs779153147 Chr.6: 42934619-42934619 on Build GRCh38
rs774004196 Chr.6: 42934527-42934527 on Build GRCh38
rs779866489 Chr.6: 42934516-42934516 on Build GRCh38
rs75557671 Chr.6: 42934548-42934548 on Build GRCh38
rs763436513 Chr.6: 42934561-42934561 on Build GRCh38
rs1008476249 Chr.6: 42934611-42934611 on Build GRCh38
rs374422440 Chr.6: 42934564-42934564 on Build GRCh38
rs756037817 Chr.6: 42934575-42934575 on Build GRCh38
rs760025708 Chr.6: 42934437-42934437 on Build GRCh38
rs1006143148 Chr.6: 42934429-42934429 on Build GRCh38
rs754771020 Chr.6: 42934608-42934608 on Build GRCh38
rs765617492 Chr.6: 42934445-42934445 on Build GRCh38
rs367900204 Chr.6: 42934631-42934631 on Build GRCh38
rs190146038 Chr.6: 42934627-42934627 on Build GRCh38
rs368438715 Chr.6: 42934623-42934623 on Build GRCh38
rs751837391 Chr.6: 42934566-42934566 on Build GRCh38
rs755208427 Chr.6: 42934547-42934547 on Build GRCh38
rs1042184488 Chr.6: 42934637-42934637 on Build GRCh38
rs746535848 Chr.6: 42934632-42934632 on Build GRCh38
rs781319933 Chr.6: 42934569-42934569 on Build GRCh38
rs201250569 Chr.6: 42934477-42934477 on Build GRCh38
rs758665561 Chr.6: 42934453-42934453 on Build GRCh38
rs553908449 Chr.6: 42934420-42934428 on Build GRCh38
rs776157697 Chr.6: 42934645-42934645 on Build GRCh38
rs757490459 Chr.6: 42934568-42934568 on Build GRCh38

View Full Product Details