Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 53048496-53048755 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA2
Entrez Gene ID
3673
Gene Name
integrin subunit alpha 2
Gene Aliases
BR, CD49B, GPIa, HPA-5, VLA-2, VLAA2
UniGene
Hs.482077
Gene Chromosome Location
Chr.5: 52989326-53094779 on Build GRCh38
Transcripts
NM_002203.3 NR_073103.1 NR_073104.1 NR_073105.1 NR_073106.1 NR_073107.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
123793 c.548A>G p.N183S Chr.5: 53048688-53048688 on Build GRCh38
305471 c.613G>C p.G205R Chr.5: 53048753-53048753 on Build GRCh38
1695768 c.512C>T p.S171F Chr.5: 53048652-53048652 on Build GRCh38
4693232 c.524T>C p.V175A Chr.5: 53048664-53048664 on Build GRCh38
5691492 c.513C>A p.S171S Chr.5: 53048653-53048653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140105624 Chr.5: 53048537-53048537 on Build GRCh38
rs750174081 Chr.5: 53048603-53048603 on Build GRCh38
rs973579586 Chr.5: 53048552-53048552 on Build GRCh38
rs963037970 Chr.5: 53048548-53048548 on Build GRCh38
rs112349018 Chr.5: 53048542-53048542 on Build GRCh38
rs776915188 Chr.5: 53048705-53048705 on Build GRCh38
rs759383464 Chr.5: 53048570-53048570 on Build GRCh38
rs555068751 Chr.5: 53048663-53048663 on Build GRCh38
rs1032422182 Chr.5: 53048716-53048716 on Build GRCh38
rs757116036 Chr.5: 53048527-53048527 on Build GRCh38
rs371749737 Chr.5: 53048594-53048594 on Build GRCh38
rs763643727 Chr.5: 53048522-53048522 on Build GRCh38
rs771303086 Chr.5: 53048674-53048674 on Build GRCh38
rs929330993 Chr.5: 53048529-53048529 on Build GRCh38
rs41268439 Chr.5: 53048626-53048626 on Build GRCh38
rs755214287 Chr.5: 53048635-53048635 on Build GRCh38
rs917545444 Chr.5: 53048661-53048661 on Build GRCh38
rs55973669 Chr.5: 53048657-53048657 on Build GRCh38
rs954624704 Chr.5: 53048537-53048537 on Build GRCh38
rs747556490 Chr.5: 53048666-53048666 on Build GRCh38
rs774574169 Chr.5: 53048630-53048630 on Build GRCh38
rs888067156 Chr.5: 53048538-53048538 on Build GRCh38
rs779046164 Chr.5: 53048652-53048652 on Build GRCh38
rs779756492 Chr.5: 53048621-53048621 on Build GRCh38
rs547370002 Chr.5: 53048575-53048575 on Build GRCh38
rs928700891 Chr.5: 53048528-53048528 on Build GRCh38
rs751132342 Chr.5: 53048525-53048525 on Build GRCh38
rs755937001 Chr.5: 53048608-53048608 on Build GRCh38
rs748243210 Chr.5: 53048653-53048653 on Build GRCh38
rs190131248 Chr.5: 53048713-53048713 on Build GRCh38

View Full Product Details