Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 151237686-151237949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHPF2
Entrez Gene ID
54480
Gene Name
chondroitin polymerizing factor 2
Gene Aliases
CSGLCA-T, CSGlcAT, ChSy-3, chPF-2
UniGene
Hs.647084
Gene Chromosome Location
Chr.7: 151232489-151238827 on Build GRCh38
Transcripts
NM_001284295.1 NM_019015.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7262283 c.1367G>A p.R456H Chr.7: 151237729-151237729 on Build GRCh38
5677938 c.1572C>G p.T524T Chr.7: 151237934-151237934 on Build GRCh38
3879434 c.1370G>C p.G457A Chr.7: 151237732-151237732 on Build GRCh38
7323481 c.1551A>T p.P517P Chr.7: 151237913-151237913 on Build GRCh38
115933 c.1400G>C p.S467T Chr.7: 151237762-151237762 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766364481 Chr.7: 151237806-151237807 on Build GRCh38
rs968620779 Chr.7: 151237834-151237834 on Build GRCh38
rs749228087 Chr.7: 151237929-151237929 on Build GRCh38
rs749772535 Chr.7: 151237737-151237737 on Build GRCh38
rs997281861 Chr.7: 151237759-151237759 on Build GRCh38
rs776144646 Chr.7: 151237758-151237758 on Build GRCh38
rs370642310 Chr.7: 151237825-151237825 on Build GRCh38
rs368760819 Chr.7: 151237721-151237721 on Build GRCh38
rs140938228 Chr.7: 151237739-151237739 on Build GRCh38
rs761507019 Chr.7: 151237874-151237874 on Build GRCh38
rs138549818 Chr.7: 151237783-151237783 on Build GRCh38
rs2487154 Chr.7: 151237889-151237889 on Build GRCh38
rs192704087 Chr.7: 151237775-151237775 on Build GRCh38
rs768451770 Chr.7: 151237847-151237847 on Build GRCh38
rs373790740 Chr.7: 151237753-151237753 on Build GRCh38
rs774127965 Chr.7: 151237873-151237873 on Build GRCh38
rs753982074 Chr.7: 151237883-151237883 on Build GRCh38
rs766374143 Chr.7: 151237799-151237799 on Build GRCh38
rs200747434 Chr.7: 151237885-151237885 on Build GRCh38
rs564817361 Chr.7: 151237807-151237807 on Build GRCh38
rs775385828 Chr.7: 151237777-151237777 on Build GRCh38
rs573622483 Chr.7: 151237746-151237746 on Build GRCh38
rs765025208 Chr.7: 151237718-151237718 on Build GRCh38
rs367935553 Chr.7: 151237824-151237824 on Build GRCh38
rs979882314 Chr.7: 151237835-151237835 on Build GRCh38
rs137963683 Chr.7: 151237720-151237720 on Build GRCh38
rs376118602 Chr.7: 151237915-151237915 on Build GRCh38
rs757542399 Chr.7: 151237902-151237902 on Build GRCh38
rs769263733 Chr.7: 151237836-151237836 on Build GRCh38
rs772776946 Chr.7: 151237743-151237743 on Build GRCh38
rs149808659 Chr.7: 151237729-151237729 on Build GRCh38
rs369590314 Chr.7: 151237912-151237912 on Build GRCh38
rs142236355 Chr.7: 151237796-151237796 on Build GRCh38
rs144589067 Chr.7: 151237741-151237741 on Build GRCh38
rs753847745 Chr.7: 151237886-151237886 on Build GRCh38
rs767776711 Chr.7: 151237727-151237727 on Build GRCh38
rs779609639 Chr.7: 151237810-151237810 on Build GRCh38
rs778146815 Chr.7: 151237822-151237822 on Build GRCh38
rs759463177 Chr.7: 151237713-151237713 on Build GRCh38
rs769785056 Chr.7: 151237773-151237773 on Build GRCh38
rs765877277 Chr.7: 151237705-151237705 on Build GRCh38
rs565541450 Chr.7: 151237707-151237707 on Build GRCh38
rs201697000 Chr.7: 151237914-151237914 on Build GRCh38
rs577213295 Chr.7: 151237738-151237738 on Build GRCh38
rs148167405 Chr.7: 151237838-151237838 on Build GRCh38
rs375138900 Chr.7: 151237851-151237851 on Build GRCh38
rs759727328 Chr.7: 151237884-151237884 on Build GRCh38
rs969808978 Chr.7: 151237819-151237821 on Build GRCh38
rs199523114 Chr.7: 151237725-151237725 on Build GRCh38
rs748578215 Chr.7: 151237740-151237740 on Build GRCh38
rs201112871 Chr.7: 151237900-151237900 on Build GRCh38
rs957281871 Chr.7: 151237793-151237793 on Build GRCh38
rs745434656 Chr.7: 151237833-151237833 on Build GRCh38
rs201586379 Chr.7: 151237844-151237844 on Build GRCh38
rs753222532 Chr.7: 151237815-151237815 on Build GRCh38
rs538931187 Chr.7: 151237731-151237731 on Build GRCh38
rs200593230 Chr.7: 151237782-151237782 on Build GRCh38
rs531048150 Chr.7: 151237791-151237791 on Build GRCh38
rs754014698 Chr.7: 151237804-151237804 on Build GRCh38
rs146815169 Chr.7: 151237819-151237819 on Build GRCh38
rs766459811 Chr.7: 151237875-151237875 on Build GRCh38
rs369576814 Chr.7: 151237927-151237927 on Build GRCh38
rs758966588 Chr.7: 151237770-151237770 on Build GRCh38
rs752021782 Chr.7: 151237901-151237901 on Build GRCh38
rs750633097 Chr.7: 151237793-151237793 on Build GRCh38

View Full Product Details