Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 57613281-57613528 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF25
Entrez Gene ID
115557
Gene Name
Rho guanine nucleotide exchange factor 25
Gene Aliases
GEFT, p63RhoGEF
UniGene
Hs.61581
Gene Chromosome Location
Chr.12: 57610180-57617245 on Build GRCh38
Transcripts
NM_001111270.2 NM_182947.3 NR_046223.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7319232 c.492C>G p.T164T Chr.12: 57613326-57613326 on Build GRCh38
6488460 c.532G>A p.E178K Chr.12: 57613446-57613446 on Build GRCh38
6875722 c.480G>A p.R160R Chr.12: 57613511-57613511 on Build GRCh38
5375713 c.472C>T p.L158L Chr.12: 57613503-57613503 on Build GRCh38
3463872 c.444G>A p.E148E Chr.12: 57613475-57613475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141337782 Chr.12: 57613366-57613366 on Build GRCh38
rs770647857 Chr.12: 57613493-57613493 on Build GRCh38
rs772921608 Chr.12: 57613363-57613363 on Build GRCh38
rs190767976 Chr.12: 57613425-57613425 on Build GRCh38
rs374927200 Chr.12: 57613407-57613407 on Build GRCh38
rs764007893 Chr.12: 57613489-57613491 on Build GRCh38
rs374323837 Chr.12: 57613421-57613421 on Build GRCh38
rs754396868 Chr.12: 57613326-57613326 on Build GRCh38
rs747710445 Chr.12: 57613356-57613356 on Build GRCh38
rs747667238 Chr.12: 57613456-57613456 on Build GRCh38
rs755771922 Chr.12: 57613461-57613461 on Build GRCh38
rs894952649 Chr.12: 57613497-57613497 on Build GRCh38
rs780840561 Chr.12: 57613350-57613350 on Build GRCh38
rs754473725 Chr.12: 57613345-57613345 on Build GRCh38
rs542498887 Chr.12: 57613499-57613499 on Build GRCh38
rs759601029 Chr.12: 57613305-57613305 on Build GRCh38
rs774269775 Chr.12: 57613399-57613399 on Build GRCh38
rs750904424 Chr.12: 57613439-57613439 on Build GRCh38
rs186523042 Chr.12: 57613400-57613400 on Build GRCh38
rs766273748 Chr.12: 57613300-57613300 on Build GRCh38
rs868770859 Chr.12: 57613452-57613452 on Build GRCh38
rs898453360 Chr.12: 57613500-57613500 on Build GRCh38
rs765908898 Chr.12: 57613427-57613427 on Build GRCh38
rs145818840 Chr.12: 57613338-57613338 on Build GRCh38
rs749148752 Chr.12: 57613477-57613477 on Build GRCh38
rs138453741 Chr.12: 57613355-57613355 on Build GRCh38
rs1002032774 Chr.12: 57613335-57613335 on Build GRCh38
rs771644007 Chr.12: 57613398-57613398 on Build GRCh38
rs777469251 Chr.12: 57613462-57613462 on Build GRCh38
rs577533598 Chr.12: 57613357-57613357 on Build GRCh38
rs770915382 Chr.12: 57613370-57613370 on Build GRCh38
rs561061153 Chr.12: 57613501-57613501 on Build GRCh38
rs143068244 Chr.12: 57613444-57613444 on Build GRCh38
rs760228384 Chr.12: 57613473-57613476 on Build GRCh38
rs767849431 Chr.12: 57613306-57613306 on Build GRCh38
rs371542216 Chr.12: 57613411-57613411 on Build GRCh38
rs746573153 Chr.12: 57613341-57613341 on Build GRCh38
rs774950194 Chr.12: 57613462-57613463 on Build GRCh38
rs61754584 Chr.12: 57613308-57613308 on Build GRCh38
rs947921217 Chr.12: 57613498-57613498 on Build GRCh38
rs140645509 Chr.12: 57613307-57613307 on Build GRCh38
rs764342424 Chr.12: 57613413-57613413 on Build GRCh38
rs201573584 Chr.12: 57613303-57613303 on Build GRCh38
rs767436829 Chr.12: 57613406-57613406 on Build GRCh38
rs544973692 Chr.12: 57613443-57613443 on Build GRCh38
rs368208007 Chr.12: 57613454-57613454 on Build GRCh38
rs757876691 Chr.12: 57613334-57613334 on Build GRCh38

View Full Product Details