Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70398443-70398712 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ST3GAL2
Entrez Gene ID
6483
Gene Name
ST3 beta-galactoside alpha-2,3-sialyltransferase 2
Gene Aliases
Gal-NAc6S, SIAT4B, ST3GALII, ST3GalA.2
UniGene
Hs.368611
Gene Chromosome Location
Chr.16: 70379435-70439088 on Build GRCh38
Transcripts
NM_006927.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6992420 c.62C>G p.S21C Chr.16: 70398469-70398469 on Build GRCh38
973012 c.16C>T p.R6W Chr.16: 70398515-70398515 on Build GRCh38
7171100 c.62C>T p.S21F Chr.16: 70398469-70398469 on Build GRCh38
5593302 c.42C>T p.F14F Chr.16: 70398489-70398489 on Build GRCh38
6455054 c.80C>T p.S27L Chr.16: 70398451-70398451 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs569746428 Chr.16: 70398609-70398609 on Build GRCh38
rs965360978 Chr.16: 70398602-70398602 on Build GRCh38
rs767852990 Chr.16: 70398543-70398543 on Build GRCh38
rs776343416 Chr.16: 70398571-70398571 on Build GRCh38
rs556433592 Chr.16: 70398684-70398684 on Build GRCh38
rs372668670 Chr.16: 70398544-70398544 on Build GRCh38
rs562210318 Chr.16: 70398546-70398546 on Build GRCh38
rs778324306 Chr.16: 70398488-70398488 on Build GRCh38
rs746821258 Chr.16: 70398503-70398503 on Build GRCh38
rs995270164 Chr.16: 70398564-70398564 on Build GRCh38
rs377720141 Chr.16: 70398507-70398507 on Build GRCh38
rs1056213428 Chr.16: 70398677-70398677 on Build GRCh38
rs764215623 Chr.16: 70398518-70398518 on Build GRCh38
rs746770139 Chr.16: 70398566-70398566 on Build GRCh38
rs529500408 Chr.16: 70398550-70398550 on Build GRCh38
rs757605200 Chr.16: 70398462-70398462 on Build GRCh38
rs371530556 Chr.16: 70398600-70398600 on Build GRCh38
rs1014762174 Chr.16: 70398685-70398685 on Build GRCh38
rs769049114 Chr.16: 70398562-70398562 on Build GRCh38
rs1001572062 Chr.16: 70398657-70398657 on Build GRCh38
rs913410163 Chr.16: 70398632-70398632 on Build GRCh38
rs376899589 Chr.16: 70398580-70398580 on Build GRCh38
rs377658212 Chr.16: 70398477-70398477 on Build GRCh38
rs771780291 Chr.16: 70398493-70398493 on Build GRCh38
rs747661710 Chr.16: 70398557-70398557 on Build GRCh38
rs373101250 Chr.16: 70398535-70398535 on Build GRCh38
rs370307749 Chr.16: 70398537-70398537 on Build GRCh38
rs367572751 Chr.16: 70398513-70398513 on Build GRCh38
rs946188376 Chr.16: 70398638-70398638 on Build GRCh38
rs768244186 Chr.16: 70398483-70398483 on Build GRCh38
rs1022256994 Chr.16: 70398673-70398673 on Build GRCh38
rs772926843 Chr.16: 70398497-70398497 on Build GRCh38
rs1028389894 Chr.16: 70398625-70398625 on Build GRCh38
rs936266271 Chr.16: 70398592-70398592 on Build GRCh38
rs539587363 Chr.16: 70398597-70398597 on Build GRCh38
rs199829873 Chr.16: 70398465-70398465 on Build GRCh38
rs766309732 Chr.16: 70398683-70398683 on Build GRCh38
rs778483764 Chr.16: 70398554-70398554 on Build GRCh38
rs769683915 Chr.16: 70398579-70398579 on Build GRCh38
rs1011459468 Chr.16: 70398547-70398547 on Build GRCh38
rs750881963 Chr.16: 70398472-70398472 on Build GRCh38
rs531316817 Chr.16: 70398533-70398533 on Build GRCh38
rs537017859 Chr.16: 70398661-70398661 on Build GRCh38
rs566261778 Chr.16: 70398563-70398563 on Build GRCh38
rs759434739 Chr.16: 70398515-70398515 on Build GRCh38
rs757990712 Chr.16: 70398558-70398558 on Build GRCh38
rs770730722 Chr.16: 70398569-70398569 on Build GRCh38
rs1048142 Chr.16: 70398627-70398627 on Build GRCh38
rs907267896 Chr.16: 70398656-70398656 on Build GRCh38
rs961888522 Chr.16: 70398689-70398689 on Build GRCh38
rs1045887976 Chr.16: 70398644-70398644 on Build GRCh38
rs1048313909 Chr.16: 70398567-70398567 on Build GRCh38
rs745808933 Chr.16: 70398576-70398576 on Build GRCh38
rs924941996 Chr.16: 70398572-70398572 on Build GRCh38
rs1004131637 Chr.16: 70398589-70398589 on Build GRCh38
rs547629531 Chr.16: 70398551-70398551 on Build GRCh38
rs371169787 Chr.16: 70398492-70398492 on Build GRCh38
rs924829655 Chr.16: 70398498-70398498 on Build GRCh38

View Full Product Details