Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134969846-134970117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPER3
Entrez Gene ID
347732
Gene Name
cation channel sperm associated 3
Gene Aliases
CACRC
UniGene
Hs.631804
Gene Chromosome Location
Chr.5: 134967906-135011707 on Build GRCh38
Transcripts
NM_178019.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
279588 c.161T>G p.I54R Chr.5: 134970001-134970001 on Build GRCh38
3850214 c.192G>A p.A64A Chr.5: 134970032-134970032 on Build GRCh38
6027122 c.204C>A p.A68A Chr.5: 134970044-134970044 on Build GRCh38
4534680 c.210G>A p.W70* Chr.5: 134970050-134970050 on Build GRCh38
3138306 c.186G>A p.S62S Chr.5: 134970026-134970026 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145333202 Chr.5: 134969989-134969989 on Build GRCh38
rs373823349 Chr.5: 134970053-134970053 on Build GRCh38
rs149256453 Chr.5: 134970093-134970093 on Build GRCh38
rs765665127 Chr.5: 134970072-134970072 on Build GRCh38
rs973678301 Chr.5: 134970039-134970039 on Build GRCh38
rs574831794 Chr.5: 134969946-134969946 on Build GRCh38
rs368208576 Chr.5: 134969927-134969927 on Build GRCh38
rs374256296 Chr.5: 134970006-134970006 on Build GRCh38
rs540871226 Chr.5: 134970044-134970044 on Build GRCh38
rs747061105 Chr.5: 134969934-134969934 on Build GRCh38
rs1015789473 Chr.5: 134970085-134970085 on Build GRCh38
rs773924046 Chr.5: 134970047-134970047 on Build GRCh38
rs12518563 Chr.5: 134969903-134969903 on Build GRCh38
rs766971508 Chr.5: 134969988-134969988 on Build GRCh38
rs765754154 Chr.5: 134969998-134969998 on Build GRCh38
rs147115565 Chr.5: 134970088-134970088 on Build GRCh38
rs756061764 Chr.5: 134970030-134970030 on Build GRCh38
rs1041530337 Chr.5: 134969959-134969959 on Build GRCh38
rs773873187 Chr.5: 134969974-134969974 on Build GRCh38
rs751605598 Chr.5: 134970015-134970015 on Build GRCh38
rs536922872 Chr.5: 134970026-134970026 on Build GRCh38
rs758393074 Chr.5: 134969921-134969921 on Build GRCh38
rs531680169 Chr.5: 134969905-134969905 on Build GRCh38
rs116119095 Chr.5: 134969956-134969956 on Build GRCh38
rs936598219 Chr.5: 134969899-134969899 on Build GRCh38
rs143170274 Chr.5: 134970063-134970063 on Build GRCh38
rs750206641 Chr.5: 134969929-134969929 on Build GRCh38
rs150251956 Chr.5: 134970025-134970025 on Build GRCh38
rs767208333 Chr.5: 134969896-134969896 on Build GRCh38
rs200787247 Chr.5: 134969961-134969961 on Build GRCh38
rs760060034 Chr.5: 134970069-134970069 on Build GRCh38
rs1052340755 Chr.5: 134969904-134969904 on Build GRCh38
rs774996033 Chr.5: 134969958-134969958 on Build GRCh38
rs574551964 Chr.5: 134970073-134970073 on Build GRCh38
rs375185027 Chr.5: 134970032-134970032 on Build GRCh38
rs757144500 Chr.5: 134969939-134969939 on Build GRCh38
rs755443631 Chr.5: 134969894-134969894 on Build GRCh38
rs753014864 Chr.5: 134970000-134970000 on Build GRCh38
rs138539609 Chr.5: 134970092-134970092 on Build GRCh38
rs1011056961 Chr.5: 134969874-134969874 on Build GRCh38
rs761253604 Chr.5: 134969976-134969976 on Build GRCh38
rs764251114 Chr.5: 134970013-134970013 on Build GRCh38
rs777265038 Chr.5: 134970065-134970065 on Build GRCh38
rs140980470 Chr.5: 134969963-134969963 on Build GRCh38
rs779835422 Chr.5: 134970031-134970031 on Build GRCh38
rs759065227 Chr.5: 134970083-134970083 on Build GRCh38
rs765524134 Chr.5: 134969915-134969915 on Build GRCh38
rs139264127 Chr.5: 134970033-134970033 on Build GRCh38
rs757373052 Chr.5: 134970023-134970023 on Build GRCh38
rs745636557 Chr.5: 134969950-134969950 on Build GRCh38
rs773884693 Chr.5: 134969981-134969981 on Build GRCh38
rs755971687 Chr.5: 134970032-134970033 on Build GRCh38

View Full Product Details