Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 101037865-101038138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC17
Entrez Gene ID
140453
Gene Name
mucin 17, cell surface associated
Gene Aliases
MUC-17, MUC-3, MUC3
UniGene
Hs.271819
Gene Chromosome Location
Chr.7: 101020083-101058859 on Build GRCh38
Transcripts
NM_001040105.1 NR_133665.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
743555 c.6706G>C p.E2236Q Chr.7: 101038122-101038122 on Build GRCh38
4389879 c.6596G>A p.R2199H Chr.7: 101038012-101038012 on Build GRCh38
1699065 c.6649G>A p.E2217K Chr.7: 101038065-101038065 on Build GRCh38
118540 c.6546A>T p.S2182S Chr.7: 101037962-101037962 on Build GRCh38
4408861 c.6474C>T p.D2158D Chr.7: 101037890-101037890 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs535379974 Chr.7: 101037980-101037980 on Build GRCh38
rs147785825 Chr.7: 101037940-101037940 on Build GRCh38
rs142126972 Chr.7: 101037985-101037985 on Build GRCh38
rs757099131 Chr.7: 101038003-101038003 on Build GRCh38
rs1037671117 Chr.7: 101037990-101037990 on Build GRCh38
rs1006145360 Chr.7: 101037978-101037978 on Build GRCh38
rs148535690 Chr.7: 101037910-101037910 on Build GRCh38
rs1046775310 Chr.7: 101037982-101037982 on Build GRCh38
rs539881448 Chr.7: 101038059-101038059 on Build GRCh38
rs148847871 Chr.7: 101038016-101038016 on Build GRCh38
rs765048960 Chr.7: 101038112-101038112 on Build GRCh38
rs773839747 Chr.7: 101037896-101037896 on Build GRCh38
rs774909654 Chr.7: 101038028-101038028 on Build GRCh38
rs147628791 Chr.7: 101038099-101038099 on Build GRCh38
rs556800891 Chr.7: 101037991-101037991 on Build GRCh38
rs763345801 Chr.7: 101037976-101037976 on Build GRCh38
rs768805511 Chr.7: 101038108-101038108 on Build GRCh38
rs752915502 Chr.7: 101037994-101037994 on Build GRCh38
rs143516283 Chr.7: 101038022-101038022 on Build GRCh38
rs557614802 Chr.7: 101038039-101038039 on Build GRCh38
rs763858746 Chr.7: 101037997-101037997 on Build GRCh38
rs372852533 Chr.7: 101037913-101037913 on Build GRCh38
rs778214077 Chr.7: 101038084-101038084 on Build GRCh38
rs28555173 Chr.7: 101037891-101037891 on Build GRCh38
rs147094151 Chr.7: 101037930-101037930 on Build GRCh38
rs139022588 Chr.7: 101038062-101038062 on Build GRCh38
rs775741685 Chr.7: 101038047-101038047 on Build GRCh38
rs752433225 Chr.7: 101037928-101037928 on Build GRCh38
rs763159692 Chr.7: 101037901-101037901 on Build GRCh38
rs745561907 Chr.7: 101037951-101037951 on Build GRCh38
rs545933092 Chr.7: 101038029-101038029 on Build GRCh38
rs778234139 Chr.7: 101037944-101037944 on Build GRCh38
rs369494380 Chr.7: 101037996-101037996 on Build GRCh38
rs778243510 Chr.7: 101038021-101038021 on Build GRCh38
rs777146561 Chr.7: 101037956-101037956 on Build GRCh38
rs144558587 Chr.7: 101038110-101038110 on Build GRCh38
rs770330569 Chr.7: 101038017-101038017 on Build GRCh38
rs141217694 Chr.7: 101038012-101038012 on Build GRCh38
rs144296263 Chr.7: 101038085-101038085 on Build GRCh38
rs763903791 Chr.7: 101038061-101038061 on Build GRCh38
rs771286219 Chr.7: 101038024-101038024 on Build GRCh38
rs750369232 Chr.7: 101038072-101038072 on Build GRCh38
rs148470778 Chr.7: 101038107-101038107 on Build GRCh38
rs767306435 Chr.7: 101037914-101037914 on Build GRCh38
rs200626179 Chr.7: 101037967-101037967 on Build GRCh38
rs149582193 Chr.7: 101038082-101038082 on Build GRCh38
rs770277203 Chr.7: 101037970-101037970 on Build GRCh38
rs146364763 Chr.7: 101038070-101038070 on Build GRCh38
rs760841706 Chr.7: 101038050-101038050 on Build GRCh38
rs746367007 Chr.7: 101038093-101038093 on Build GRCh38
rs756844699 Chr.7: 101037941-101037941 on Build GRCh38
rs140606802 Chr.7: 101038109-101038109 on Build GRCh38
rs751830299 Chr.7: 101037911-101037911 on Build GRCh38
rs377045493 Chr.7: 101037938-101037938 on Build GRCh38
rs144624588 Chr.7: 101038092-101038092 on Build GRCh38
rs772111228 Chr.7: 101038042-101038042 on Build GRCh38
rs145280834 Chr.7: 101037955-101037955 on Build GRCh38
rs572884234 Chr.7: 101038046-101038046 on Build GRCh38
rs772166358 Chr.7: 101038096-101038096 on Build GRCh38
rs747231852 Chr.7: 101038106-101038106 on Build GRCh38
rs528861361 Chr.7: 101038077-101038077 on Build GRCh38
rs765186163 Chr.7: 101038073-101038083 on Build GRCh38
rs1016884814 Chr.7: 101038044-101038044 on Build GRCh38
rs551587759 Chr.7: 101037890-101037890 on Build GRCh38
rs148707760 Chr.7: 101038086-101038086 on Build GRCh38
rs11979706 Chr.7: 101038078-101038078 on Build GRCh38
rs753840014 Chr.7: 101037998-101037998 on Build GRCh38
rs533982168 Chr.7: 101037892-101037892 on Build GRCh38
rs778488002 Chr.7: 101038004-101038004 on Build GRCh38
rs773896553 Chr.7: 101037975-101037975 on Build GRCh38
rs201412788 Chr.7: 101038036-101038036 on Build GRCh38
rs142097516 Chr.7: 101038064-101038064 on Build GRCh38
rs948252190 Chr.7: 101037949-101037949 on Build GRCh38
rs373695722 Chr.7: 101038011-101038011 on Build GRCh38
rs771075843 Chr.7: 101037979-101037979 on Build GRCh38

View Full Product Details