Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 56060206-56060477 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCT6A
Entrez Gene ID
908
Gene Name
chaperonin containing TCP1 subunit 6A
Gene Aliases
CCT-zeta, CCT-zeta-1, CCT6, Cctz, HTR3, MoDP-2, TCP-1-zeta, TCP20, TCPZ, TTCP20
UniGene
Hs.82916
Gene Chromosome Location
Chr.7: 56051685-56063989 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3 NR_002957.1

Gene Symbol
SNORA15
Entrez Gene ID
677803
Gene Name
small nucleolar RNA, H/ACA box 15
Gene Aliases
ACA15, SNORA15A
UniGene
Hs.689706
Gene Chromosome Location
Chr.7: 56060470-56060602 on Build GRCh38
Transcripts
NM_001009186.1 NM_001762.3 NR_002957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6873449 c.1187G>A p.R396K Chr.7: 56060390-56060390 on Build GRCh38
1090946 c.1072G>C p.E358Q Chr.7: 56060275-56060275 on Build GRCh38
1090947 c.1109G>A p.R370H Chr.7: 56060312-56060312 on Build GRCh38
7100351 c.1156C>G p.Q386E Chr.7: 56060359-56060359 on Build GRCh38
6353280 c.1204A>G p.I402V Chr.7: 56060407-56060407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765242053 Chr.7: 56060429-56060429 on Build GRCh38
rs779443890 Chr.7: 56060239-56060239 on Build GRCh38
rs750876408 Chr.7: 56060349-56060349 on Build GRCh38
rs370258018 Chr.7: 56060458-56060458 on Build GRCh38
rs370756693 Chr.7: 56060415-56060415 on Build GRCh38
rs755711014 Chr.7: 56060361-56060361 on Build GRCh38
rs201280999 Chr.7: 56060353-56060353 on Build GRCh38
rs142501344 Chr.7: 56060405-56060405 on Build GRCh38
rs762258897 Chr.7: 56060250-56060251 on Build GRCh38
rs398004857 Chr.7: 56060258-56060258 on Build GRCh38
rs750504814 Chr.7: 56060437-56060437 on Build GRCh38
rs71015173 Chr.7: 56060253-56060253 on Build GRCh38
rs750543525 Chr.7: 56060443-56060444 on Build GRCh38
rs747738564 Chr.7: 56060283-56060283 on Build GRCh38
rs748720687 Chr.7: 56060373-56060373 on Build GRCh38
rs772595363 Chr.7: 56060286-56060286 on Build GRCh38
rs781441719 Chr.7: 56060457-56060457 on Build GRCh38
rs111336486 Chr.7: 56060354-56060354 on Build GRCh38
rs763444281 Chr.7: 56060439-56060439 on Build GRCh38
rs751291094 Chr.7: 56060408-56060408 on Build GRCh38
rs760744124 Chr.7: 56060311-56060311 on Build GRCh38
rs747476832 Chr.7: 56060358-56060358 on Build GRCh38
rs770754886 Chr.7: 56060290-56060290 on Build GRCh38
rs372118584 Chr.7: 56060264-56060264 on Build GRCh38
rs773199376 Chr.7: 56060259-56060259 on Build GRCh38
rs148413036 Chr.7: 56060241-56060241 on Build GRCh38
rs777534496 Chr.7: 56060371-56060371 on Build GRCh38
rs141113289 Chr.7: 56060397-56060397 on Build GRCh38
rs774475133 Chr.7: 56060296-56060296 on Build GRCh38
rs778588911 Chr.7: 56060375-56060375 on Build GRCh38
rs757397183 Chr.7: 56060333-56060333 on Build GRCh38
rs770524855 Chr.7: 56060374-56060374 on Build GRCh38
rs765327797 Chr.7: 56060342-56060342 on Build GRCh38
rs534008553 Chr.7: 56060382-56060382 on Build GRCh38
rs536595674 Chr.7: 56060235-56060235 on Build GRCh38
rs767372493 Chr.7: 56060300-56060300 on Build GRCh38
rs1005532616 Chr.7: 56060410-56060410 on Build GRCh38
rs769489074 Chr.7: 56060285-56060285 on Build GRCh38
rs151087192 Chr.7: 56060364-56060364 on Build GRCh38
rs759578820 Chr.7: 56060298-56060298 on Build GRCh38
rs755411374 Chr.7: 56060448-56060448 on Build GRCh38
rs764432664 Chr.7: 56060312-56060312 on Build GRCh38
rs780415594 Chr.7: 56060270-56060270 on Build GRCh38
rs780327639 Chr.7: 56060356-56060356 on Build GRCh38
rs762856914 Chr.7: 56060306-56060306 on Build GRCh38
rs760725069 Chr.7: 56060446-56060447 on Build GRCh38
rs371562349 Chr.7: 56060399-56060399 on Build GRCh38
rs566507599 Chr.7: 56060330-56060330 on Build GRCh38
rs766930562 Chr.7: 56060445-56060445 on Build GRCh38
rs192970041 Chr.7: 56060407-56060407 on Build GRCh38

View Full Product Details