Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 49031482-49031711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TFE3
Entrez Gene ID
7030
Gene Name
transcription factor binding to IGHM enhancer 3
Gene Aliases
RCCP2, RCCX1, TFEA, bHLHe33
UniGene
Hs.730740
Gene Chromosome Location
Chr.X: 49028726-49043517 on Build GRCh38
Transcripts
NM_001282142.1 NM_006521.5

SNP Detail

SNP ID Chromosome Location
rs967351054 Chr.X: 49031609-49031609 on Build GRCh38
rs782039733 Chr.X: 49031617-49031617 on Build GRCh38
rs782357804 Chr.X: 49031593-49031593 on Build GRCh38
rs782282280 Chr.X: 49031549-49031549 on Build GRCh38
rs782213833 Chr.X: 49031573-49031573 on Build GRCh38
rs781999503 Chr.X: 49031569-49031569 on Build GRCh38
rs782683383 Chr.X: 49031553-49031553 on Build GRCh38
rs781946230 Chr.X: 49031597-49031597 on Build GRCh38

View Full Product Details