Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8044778-8045051 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALOX15B
Entrez Gene ID
247
Gene Name
arachidonate 15-lipoxygenase, type B
Gene Aliases
15-LOX-2
UniGene
Hs.111256
Gene Chromosome Location
Chr.17: 8039040-8049134 on Build GRCh38
Transcripts
NM_001039130.1 NM_001039131.1 NM_001141.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1182578 c.767A>G p.Y256C Chr.17: 8044919-8044919 on Build GRCh38
148184 c.705C>T p.D235D Chr.17: 8044857-8044857 on Build GRCh38
5833468 c.810_811delAG p.L272fs*9 Chr.17: 8044962-8044963 on Build GRCh38
1182577 c.706G>A p.A236T Chr.17: 8044858-8044858 on Build GRCh38
4661201 c.714C>A p.F238L Chr.17: 8044866-8044866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776057927 Chr.17: 8045009-8045009 on Build GRCh38
rs200024960 Chr.17: 8045007-8045007 on Build GRCh38
rs760319119 Chr.17: 8045024-8045024 on Build GRCh38
rs1012306959 Chr.17: 8044819-8044819 on Build GRCh38
rs540243886 Chr.17: 8044805-8044805 on Build GRCh38
rs563147555 Chr.17: 8044933-8044933 on Build GRCh38
rs141534086 Chr.17: 8044921-8044921 on Build GRCh38
rs1032839338 Chr.17: 8044841-8044841 on Build GRCh38
rs777839317 Chr.17: 8044958-8044958 on Build GRCh38
rs990713839 Chr.17: 8044995-8044995 on Build GRCh38
rs757201622 Chr.17: 8044866-8044866 on Build GRCh38
rs529020211 Chr.17: 8044809-8044809 on Build GRCh38
rs375708086 Chr.17: 8044867-8044867 on Build GRCh38
rs147889634 Chr.17: 8044909-8044909 on Build GRCh38
rs748409515 Chr.17: 8044883-8044883 on Build GRCh38
rs771343490 Chr.17: 8044897-8044897 on Build GRCh38
rs199761192 Chr.17: 8044802-8044802 on Build GRCh38
rs959262356 Chr.17: 8044982-8044982 on Build GRCh38
rs146184262 Chr.17: 8044975-8044975 on Build GRCh38
rs202104819 Chr.17: 8044822-8044822 on Build GRCh38
rs533126418 Chr.17: 8044833-8044833 on Build GRCh38
rs866474395 Chr.17: 8044945-8044945 on Build GRCh38
rs769911219 Chr.17: 8044885-8044885 on Build GRCh38
rs757153840 Chr.17: 8044938-8044938 on Build GRCh38
rs1049246703 Chr.17: 8044925-8044925 on Build GRCh38
rs750970244 Chr.17: 8044810-8044810 on Build GRCh38
rs183068946 Chr.17: 8044814-8044814 on Build GRCh38
rs994966995 Chr.17: 8044932-8044932 on Build GRCh38
rs561621979 Chr.17: 8044807-8044807 on Build GRCh38
rs756302383 Chr.17: 8044952-8044952 on Build GRCh38
rs749744179 Chr.17: 8044895-8044895 on Build GRCh38
rs753813375 Chr.17: 8044858-8044858 on Build GRCh38
rs201480696 Chr.17: 8044939-8044939 on Build GRCh38
rs773738143 Chr.17: 8044830-8044830 on Build GRCh38
rs969827001 Chr.17: 8044829-8044829 on Build GRCh38
rs749355461 Chr.17: 8044974-8044974 on Build GRCh38
rs149155475 Chr.17: 8044900-8044900 on Build GRCh38
rs755154224 Chr.17: 8044871-8044871 on Build GRCh38
rs567291621 Chr.17: 8044935-8044935 on Build GRCh38
rs890032833 Chr.17: 8044879-8044879 on Build GRCh38
rs776626203 Chr.17: 8044818-8044818 on Build GRCh38
rs768039134 Chr.17: 8044908-8044908 on Build GRCh38
rs759189692 Chr.17: 8044853-8044853 on Build GRCh38
rs774972084 Chr.17: 8044979-8044979 on Build GRCh38
rs1012206869 Chr.17: 8044990-8044990 on Build GRCh38
rs779889902 Chr.17: 8044950-8044950 on Build GRCh38
rs6503070 Chr.17: 8044857-8044857 on Build GRCh38
rs746421707 Chr.17: 8045003-8045003 on Build GRCh38
rs758469928 Chr.17: 8044925-8044931 on Build GRCh38
rs981965839 Chr.17: 8044953-8044953 on Build GRCh38
rs142322486 Chr.17: 8044907-8044907 on Build GRCh38
rs781414757 Chr.17: 8044876-8044876 on Build GRCh38
rs760315762 Chr.17: 8044910-8044910 on Build GRCh38
rs367867264 Chr.17: 8044951-8044951 on Build GRCh38
rs552734608 Chr.17: 8044803-8044803 on Build GRCh38
rs143883230 Chr.17: 8044834-8044834 on Build GRCh38
rs768199728 Chr.17: 8045031-8045031 on Build GRCh38

View Full Product Details