Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 58098676-58098949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLNB
Entrez Gene ID
2317
Gene Name
filamin B
Gene Aliases
ABP-278, ABP-280, AOI, FH1, FLN-B, FLN1L, LRS1, SCT, TABP, TAP
UniGene
Hs.476448
Gene Chromosome Location
Chr.3: 58008400-58172255 on Build GRCh38
Transcripts
NM_001164317.1 NM_001164318.1 NM_001164319.1 NM_001457.3 XM_005264978.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5198246 c.1202C>A p.T401N Chr.3: 58098765-58098765 on Build GRCh38
3596737 c.1179G>A p.V393V Chr.3: 58098742-58098742 on Build GRCh38
6544491 c.1158G>T p.V386V Chr.3: 58098721-58098721 on Build GRCh38
1593982 c.1327G>A p.V443I Chr.3: 58098890-58098890 on Build GRCh38
3916521 c.1312C>T p.P438S Chr.3: 58098875-58098875 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs192933125 Chr.3: 58098862-58098862 on Build GRCh38
rs762223685 Chr.3: 58098757-58098757 on Build GRCh38
rs768749018 Chr.3: 58098895-58098895 on Build GRCh38
rs774917649 Chr.3: 58098820-58098820 on Build GRCh38
rs762124233 Chr.3: 58098835-58098835 on Build GRCh38
rs146583950 Chr.3: 58098812-58098812 on Build GRCh38
rs368729696 Chr.3: 58098711-58098711 on Build GRCh38
rs764760156 Chr.3: 58098767-58098767 on Build GRCh38
rs777753260 Chr.3: 58098776-58098776 on Build GRCh38
rs767390595 Chr.3: 58098926-58098927 on Build GRCh38
rs765983874 Chr.3: 58098852-58098854 on Build GRCh38
rs927026973 Chr.3: 58098810-58098810 on Build GRCh38
rs149133353 Chr.3: 58098849-58098849 on Build GRCh38
rs367842487 Chr.3: 58098779-58098779 on Build GRCh38
rs145673747 Chr.3: 58098759-58098759 on Build GRCh38
rs200902568 Chr.3: 58098890-58098890 on Build GRCh38
rs148507053 Chr.3: 58098766-58098766 on Build GRCh38
rs763583078 Chr.3: 58098734-58098734 on Build GRCh38
rs751440006 Chr.3: 58098923-58098924 on Build GRCh38
rs781617769 Chr.3: 58098789-58098789 on Build GRCh38
rs147854989 Chr.3: 58098853-58098853 on Build GRCh38
rs572259451 Chr.3: 58098841-58098841 on Build GRCh38
rs746116976 Chr.3: 58098800-58098800 on Build GRCh38
rs755016228 Chr.3: 58098924-58098925 on Build GRCh38
rs779336330 Chr.3: 58098899-58098899 on Build GRCh38
rs368128453 Chr.3: 58098889-58098889 on Build GRCh38
rs752136994 Chr.3: 58098773-58098773 on Build GRCh38
rs376909618 Chr.3: 58098838-58098838 on Build GRCh38
rs775659462 Chr.3: 58098729-58098729 on Build GRCh38
rs150637036 Chr.3: 58098775-58098775 on Build GRCh38
rs775714153 Chr.3: 58098806-58098806 on Build GRCh38
rs751496341 Chr.3: 58098860-58098860 on Build GRCh38
rs763982695 Chr.3: 58098856-58098856 on Build GRCh38
rs774588966 Chr.3: 58098749-58098749 on Build GRCh38
rs903845836 Chr.3: 58098878-58098878 on Build GRCh38
rs755517689 Chr.3: 58098830-58098830 on Build GRCh38
rs982545574 Chr.3: 58098887-58098887 on Build GRCh38
rs1042432208 Chr.3: 58098863-58098863 on Build GRCh38
rs889844977 Chr.3: 58098915-58098915 on Build GRCh38
rs377651340 Chr.3: 58098778-58098778 on Build GRCh38
rs753455911 Chr.3: 58098762-58098762 on Build GRCh38
rs745987694 Chr.3: 58098713-58098713 on Build GRCh38
rs773581431 Chr.3: 58098924-58098924 on Build GRCh38
rs375562059 Chr.3: 58098912-58098912 on Build GRCh38
rs371999821 Chr.3: 58098807-58098807 on Build GRCh38
rs764813590 Chr.3: 58098842-58098842 on Build GRCh38
rs764591872 Chr.3: 58098741-58098741 on Build GRCh38
rs975445098 Chr.3: 58098923-58098923 on Build GRCh38
rs771120650 Chr.3: 58098707-58098707 on Build GRCh38
rs371126835 Chr.3: 58098717-58098717 on Build GRCh38
rs781162510 Chr.3: 58098866-58098866 on Build GRCh38
rs745588918 Chr.3: 58098884-58098884 on Build GRCh38
rs770097697 Chr.3: 58098804-58098804 on Build GRCh38
rs376416099 Chr.3: 58098703-58098703 on Build GRCh38
rs892623277 Chr.3: 58098719-58098719 on Build GRCh38
rs772502797 Chr.3: 58098920-58098920 on Build GRCh38

View Full Product Details