Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43539946-43540195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB6
Entrez Gene ID
64800
Gene Name
EF-hand calcium binding domain 6
Gene Aliases
DJBP, HSCBCIP1, dJ185D5.1
UniGene
Hs.658996
Gene Chromosome Location
Chr.22: 43528549-43812403 on Build GRCh38
Transcripts
NM_022785.3 NM_198856.2 XM_005261704.2 XM_005261705.3 XM_011530316.1 XM_011530317.2 XM_011530318.2 XM_011530319.2 XM_011530320.2 XM_011530321.2 XM_011530322.2 XM_011530323.2 XM_011530325.2 XM_011530327.2 XM_011530329.1 XM_017028910.1 XM_017028911.1 XM_017028912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4386365 c.3824T>C p.L1275P Chr.22: 43540182-43540182 on Build GRCh38
373096 c.3846G>C p.L1282L Chr.22: 43540160-43540160 on Build GRCh38
5393796 c.3840G>A p.Q1280Q Chr.22: 43540166-43540166 on Build GRCh38
5393795 c.3841G>A p.E1281K Chr.22: 43540165-43540165 on Build GRCh38
3912933 c.3833C>T p.P1278L Chr.22: 43540173-43540173 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs918356904 Chr.22: 43539999-43540000 on Build GRCh38
rs1015862820 Chr.22: 43540000-43540000 on Build GRCh38
rs951857911 Chr.22: 43540089-43540089 on Build GRCh38
rs559316817 Chr.22: 43540096-43540096 on Build GRCh38
rs373112142 Chr.22: 43540162-43540162 on Build GRCh38
rs879059227 Chr.22: 43540060-43540060 on Build GRCh38
rs772673519 Chr.22: 43540095-43540095 on Build GRCh38
rs941230751 Chr.22: 43539969-43539969 on Build GRCh38
rs371386775 Chr.22: 43540013-43540013 on Build GRCh38
rs773822530 Chr.22: 43540143-43540143 on Build GRCh38
rs530469013 Chr.22: 43540044-43540044 on Build GRCh38
rs757422578 Chr.22: 43540086-43540086 on Build GRCh38
rs112538076 Chr.22: 43540051-43540051 on Build GRCh38
rs753936807 Chr.22: 43540081-43540081 on Build GRCh38
rs368908026 Chr.22: 43540142-43540142 on Build GRCh38
rs944388116 Chr.22: 43540122-43540122 on Build GRCh38
rs377450461 Chr.22: 43540167-43540167 on Build GRCh38
rs924532150 Chr.22: 43540135-43540135 on Build GRCh38
rs772930681 Chr.22: 43540129-43540129 on Build GRCh38
rs972441030 Chr.22: 43539990-43539990 on Build GRCh38
rs780612474 Chr.22: 43540097-43540097 on Build GRCh38
rs747422256 Chr.22: 43540105-43540105 on Build GRCh38
rs938993426 Chr.22: 43540008-43540008 on Build GRCh38
rs372744887 Chr.22: 43540123-43540123 on Build GRCh38
rs904010720 Chr.22: 43540076-43540076 on Build GRCh38
rs778091628 Chr.22: 43540078-43540078 on Build GRCh38
rs371189027 Chr.22: 43540169-43540169 on Build GRCh38
rs779120010 Chr.22: 43540093-43540093 on Build GRCh38
rs759521381 Chr.22: 43540149-43540149 on Build GRCh38
rs186071413 Chr.22: 43539991-43539991 on Build GRCh38
rs762500497 Chr.22: 43540139-43540139 on Build GRCh38
rs746442051 Chr.22: 43540094-43540094 on Build GRCh38
rs536611263 Chr.22: 43539980-43539980 on Build GRCh38

View Full Product Details