Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001080974.2 | NP_001074443.1 | 5 | 367 | |
NM_025618.3 | NP_079894.2 | 5 | 407 | ||
GenBank mRNA | AK008404.1 | - | 5 | 359 | |
AK008783.1 | - | 5 | 408 | ||
AK008970.1 | - | 5 | 356 | ||
AK134747.1 | - | 5 | 369 | ||
AK146071.1 | - | 5 | 364 | ||
AK146911.1 | - | 5 | 348 | ||
AK168572.1 | - | 5 | 348 | ||
AK168792.1 | - | 5 | 348 | ||
AK168900.1 | - | 5 | 339 | ||
BC065790.1 | - | 5 | 334 | 948616 | |
BY708689.1 | - | 5 | 405 |