Gene Transcripts

Gene Symbol
Epn3
Entrez Gene ID
71889
Gene Name
epsin 3
Gene Aliases
2310022G12Rik
Chromosome Location
Chr. 11: 94489599 - 94499974 on Build GRCm38
UniGene ID
Mm.248620
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027984.3 NP_082260.1 8, 9 1769
GenBank mRNA AK009469.1 - 7, 8 1149
AK158031.1 - 7, 8 1647
BC016454.1 - 8, 9 1718 4217221

View Full Product Details