Gene Transcripts

Gene Symbol
Lmnb2
Entrez Gene ID
16907
Gene Name
lamin B2
Gene Aliases
Chromosome Location
Chr. 10: 80901363 - 80918245 on Build GRCm38
UniGene ID
Mm.7362
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010722.5 NP_034852.2 3, 4 582
GenBank mRNA BC042430.1 - 3, 4 587
BC051985.1 - 3, 4 558 5695459
X54098.1 - 3, 4 541

View Full Product Details