Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153143.4 | NP_694783.1 | 1 | 795 | |
GenBank mRNA | AF465352.1 | - | 1 | 762 | |
AK039558.1 | - | 1217 | |||
AK161602.1 | - | 1 | 790 | ||
AK170258.1 | - | 1 | 689 | ||
BC052502.1 | - | 1 | 43 | 5119264 | |
BC052734.1 | - | 1 | 746 | 6816736 | |
BC063052.1 | - | 1 | 746 |