Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144526.3 | NP_653109.2 | 3 | 407 | |
GenBank mRNA | AK011345.1 | - | 3 | 406 | |
AK049710.1 | - | 3 | 408 | ||
AK078429.1 | - | 3 | 408 | ||
BC020115.1 | - | 3 | 393 | 3988382 | |
BC029115.1 | - | 3 | 393 | 3983148 |