Gene Transcripts
- Gene Symbol
- Cd44
- Entrez Gene ID
- 12505
- Gene Name
- CD44 antigen
- Gene Aliases
- AU023126, AW121933, AW146109, HERMES, Ly-24, Pgp-1
- Chromosome Location
- Chr. 2: 102811141 - 102901669 on Build GRCm38
- UniGene ID
- Mm.423621
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039150.1 |
NP_001034239.1
|
11, 12 |
1701 |
|
NM_001039151.1 |
NP_001034240.1
|
6, 7 |
1056 |
|
NM_001177785.1 |
NP_001171256.1
|
13, 14 |
1932 |
|
NM_001177786.1 |
NP_001171257.1
|
9, 10 |
1455 |
|
NM_001177787.1 |
NP_001171258.1
|
8, 9 |
1353 |
|
NM_009851.2 |
NP_033981.2
|
16, 17 |
2301 |
|
GenBank mRNA |
AJ251594.1 |
- |
16, 17 |
2088 |
|
AK045226.1 |
- |
6, 7 |
902 |
|
AK144125.1 |
- |
6, 7 |
859 |
|
AK152099.1 |
- |
6, 7 |
852 |
|
AK154411.1 |
- |
6, 7 |
847 |
|
AK162720.1 |
- |
6, 7 |
853 |
|
AK166320.1 |
- |
6, 7 |
841 |
|
BC005676.1 |
- |
6, 7 |
835 |
|
BC051388.1 |
- |
11, 12 |
1531 |
4910789 |
CT010324.1 |
- |
11, 12 |
1382 |
|
J05163.1 |
- |
6, 7 |
718 |
|
M27129.1 |
- |
6, 7 |
763 |
|
M27130.1 |
- |
6, 7 |
769 |
|
M30655.1 |
- |
6, 7 |
837 |
|
X66081.1 |
- |
8, 9 |
1037 |
|
X66082.1 |
- |
9, 10 |
1139 |
|
X66083.1 |
- |
11, 12 |
1385 |
|
X66084.1 |
- |
13, 14 |
1616 |
|
View Full Product Details