Gene Transcripts

Gene Symbol
Daf2
Entrez Gene ID
13137
Gene Name
decay accelerating factor 2
Gene Aliases
AI323748, Cd55b, Daf, Daf-TM, TM-DAF
Chromosome Location
Chr. 1: 130388528 - 130423037 on Build GRCm38
UniGene ID
Mm.436759
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007827.2 NP_031853.2 6 774
GenBank mRNA BC115954.1 - 6 774
L41365.1 - 6 783

View Full Product Details