Gene Transcripts

Gene Symbol
Fbxw16
Entrez Gene ID
320083
Gene Name
F-box and WD-40 domain protein 16
Gene Aliases
7420402K12Rik
Chromosome Location
Chr. 9: 109432318 - 109449140 on Build GRCm38
UniGene ID
Mm.304296
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177070.3 NP_796044.2 6 772
GenBank mRNA AK078661.1 - 6 774
BC100319.1 - 6 756 30940137
DV662133.1 - 1 49

View Full Product Details