Gene Transcripts

Gene Symbol
Snhg17
Entrez Gene ID
68108
Gene Name
small nucleolar RNA host gene 17
Gene Aliases
9430008C03Rik
Chromosome Location
Chr. 2: 158353700 - 158361522 on Build GRCm38
UniGene ID
Mm.119198
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK007971.1 - 2, 3 236
AK040893.1 - 2, 3 227
AK046044.1 - 2, 3 336
AK079109.1 - 2, 3 236

View Full Product Details