Gene Transcripts

Gene Symbol
Fance
Entrez Gene ID
72775
Gene Name
Fanconi anemia, complementation group E
Gene Aliases
2810451D06Rik, AI415634, AW209126
Chromosome Location
Chr. 17: 28313530 - 28326574 on Build GRCm38
UniGene ID
Mm.201859
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163819.1 NP_001157291.1 2 682
NM_001163820.1 NP_001157292.1 2 682
GenBank mRNA AK013325.1 - 2 645
AK135017.1 - 3 586
AK172283.1 - 2 666
AK213002.1 - 1 88
BC016538.1 - 2 640
BC039987.1 - 2 645
BY740269.1 - 2 666

View Full Product Details