Gene Transcripts

Gene Symbol
Notch1
Entrez Gene ID
25496
Gene Name
notch 1
Gene Aliases
NOTCH, TAN1
Chromosome Location
Chr. 3: 3905562 - 3951015 on Build Rnor_6.0
UniGene ID
Rn.25046
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105721.1 NP_001099191.1 17 2939
GenBank mRNA X57405.1 - 17 2953

View Full Product Details