Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001106249.2 | NP_001099719.2 | 8 | 1531 | |
XM_006228946.2 | XP_006229008.1 | 8 | 1647 | ||
XM_008759327.1 | XP_008757549.1 | 8 | 1622 | ||
XM_008759328.1 | XP_008757550.1 | 8 | 1619 |