Gene Transcripts

Gene Symbol
Abca12
Entrez Gene ID
301482
Gene Name
ATP-binding cassette, subfamily A (ABC1), member 12
Gene Aliases
Chromosome Location
Chr. 9: 78521576 - 78693403 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_237242.9 XP_237242.6 28 4380

View Full Product Details