Gene Transcripts

Gene Symbol
Adamtsl2
Entrez Gene ID
311827
Gene Name
ADAMTS-like 2
Gene Aliases
RGD1305459
Chromosome Location
Chr. 3: 5624474 - 5654890 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_231125.7 XP_231125.3 12 1936

View Full Product Details