Gene Transcripts

Gene Symbol
Gulo
Entrez Gene ID
60671
Gene Name
gulonolactone (L-) oxidase
Gene Aliases
Chromosome Location
Chr. 15: 42671206 - 42693711 on Build Rnor_6.0
UniGene ID
Rn.115212
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022220.2 NP_071556.2 9 853
XM_006252146.2 XP_006252208.1 9 994
GenBank mRNA BC089803.1 - 9 853
FQ219140.1 - 9 878

View Full Product Details