Gene Transcripts

Gene Symbol
S100g
Entrez Gene ID
24249
Gene Name
S100 calcium binding protein G
Gene Aliases
Cabp, Calb3, Cbpi, Rncalbd9
Chromosome Location
Chr. X: 33442811 - 33445714 on Build Rnor_6.0
UniGene ID
Rn.9974
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012521.2 NP_036653.1 2, 3 194
GenBank mRNA BC059153.1 - 2, 3 198
J02954.1 - 2, 3 195

View Full Product Details