Gene Transcripts

Gene Symbol
Smcp
Entrez Gene ID
24899
Gene Name
sperm mitochondria-associated cysteine-rich protein
Gene Aliases
Mcs, Mcsp
Chromosome Location
Chr. 2: 192775425 - 192780631 on Build Rnor_6.0
UniGene ID
Rn.10492
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031536.2 NP_113724.1 2 484
GenBank mRNA BC078795.1 - 2 395
BF415595.1 - 1 380
U48702.1 - 2 490
X87883.1 - 2 476

View Full Product Details