Gene Transcripts

Gene Symbol
Mitf
Entrez Gene ID
25094
Gene Name
microphthalmia-associated transcription factor
Gene Aliases
Chromosome Location
Chr. 4: 130342624 - 130422735 on Build Rnor_6.0
UniGene ID
Rn.31427
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF029886.1 - 2 124

View Full Product Details