Gene Transcripts

Gene Symbol
Mal
Entrez Gene ID
25263
Gene Name
mal, T-cell differentiation protein
Gene Aliases
MALGENE
Chromosome Location
Chr. 3: 120209647 - 120233655 on Build Rnor_6.0
UniGene ID
Rn.10174
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012798.1 NP_036930.1 2 295
GenBank mRNA BC085755.1 - 2 256
U31367.1 - 2 295
X82557.1 - 2 266

View Full Product Details