Gene Transcripts

Gene Symbol
LOC498705
Entrez Gene ID
498705
Gene Name
hypothetical protein LOC498705
Gene Aliases
Chromosome Location
Chr. 17: 9972796 - 9982609 on Build Rnor_6.0
UniGene ID
Rn.123871
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001047934.1 NP_001041399.1 1, 2 132
GenBank mRNA AY539928.1 - 1, 2 132

View Full Product Details