Gene Transcripts

Gene Symbol
Ric3
Entrez Gene ID
687147
Gene Name
RIC3 acetylcholine receptor chaperone
Gene Aliases
Chromosome Location
Chr. 1: 173628281 - 173682274 on Build Rnor_6.0
UniGene ID
Rn.219078
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001115045.3 NP_001108517.1 5 577
NM_001277069.1 NP_001263998.1 5 574
GenBank mRNA AM422212.1 - 5 574
AM422213.1 - 5 577

View Full Product Details