Gene Transcripts

Gene Symbol
Morc2b
Entrez Gene ID
240069
Gene Name
microrchidia 2B
Gene Aliases
4932411A10Rik, Tce6
Chromosome Location
Chr.17: 33135588 - 33150499 on Build GRCm38
Uni Gene
Mm.157765
Interrogated Sequence Translated Protein
RefSeq NM_177719.4 NP_808387.2
GenBank mRNA AK077016.1 -
BC120896.1 -
BC120897.1 -

View Full Product Details